Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24364101-24364604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT20HL1
Entrez Gene ID
100130302
Gene Name
SPT20 homolog, SAGA complex component-like 1
Gene Aliases
FAM48B1, SPT20L
UniGene
Hs.717243
Gene Chromosome Location
Chr.X: 24362761-24365424 on Build GRCh38
Transcripts
NM_001136234.1

SNP Detail

SNP ID Chromosome Location
rs201827126 Chr.X: 24364256-24364256 on Build GRCh38
rs761086669 Chr.X: 24364373-24364373 on Build GRCh38
rs768988575 Chr.X: 24364318-24364318 on Build GRCh38
rs748979416 Chr.X: 24364495-24364495 on Build GRCh38
rs745313631 Chr.X: 24364516-24364516 on Build GRCh38
rs772857345 Chr.X: 24364240-24364240 on Build GRCh38
rs776061720 Chr.X: 24364291-24364291 on Build GRCh38
rs374642364 Chr.X: 24364547-24364547 on Build GRCh38
rs772476871 Chr.X: 24364122-24364122 on Build GRCh38
rs369594520 Chr.X: 24364443-24364443 on Build GRCh38
rs757250153 Chr.X: 24364521-24364522 on Build GRCh38
rs780987993 Chr.X: 24364355-24364371 on Build GRCh38
rs773674309 Chr.X: 24364398-24364398 on Build GRCh38
rs757084904 Chr.X: 24364353-24364373 on Build GRCh38
rs765320766 Chr.X: 24364383-24364383 on Build GRCh38
rs376276028 Chr.X: 24364554-24364554 on Build GRCh38
rs113091397 Chr.X: 24364312-24364312 on Build GRCh38
rs762506994 Chr.X: 24364170-24364170 on Build GRCh38
rs767696352 Chr.X: 24364242-24364256 on Build GRCh38
rs750621201 Chr.X: 24364272-24364272 on Build GRCh38
rs758306853 Chr.X: 24364257-24364262 on Build GRCh38
rs190025879 Chr.X: 24364381-24364381 on Build GRCh38
rs756589933 Chr.X: 24364474-24364474 on Build GRCh38
rs751560001 Chr.X: 24364181-24364181 on Build GRCh38
rs773674494 Chr.X: 24364385-24364390 on Build GRCh38
rs867801270 Chr.X: 24364309-24364309 on Build GRCh38
rs902434866 Chr.X: 24364408-24364425 on Build GRCh38
rs761098695 Chr.X: 24364408-24364409 on Build GRCh38
rs762422141 Chr.X: 24364307-24364312 on Build GRCh38
rs775103485 Chr.X: 24364375-24364381 on Build GRCh38
rs763934492 Chr.X: 24364544-24364544 on Build GRCh38
rs750710743 Chr.X: 24364452-24364452 on Build GRCh38
rs764430993 Chr.X: 24364375-24364375 on Build GRCh38
rs772733261 Chr.X: 24364279-24364279 on Build GRCh38
rs868346878 Chr.X: 24364303-24364303 on Build GRCh38
rs866630199 Chr.X: 24364422-24364422 on Build GRCh38
rs777743037 Chr.X: 24364257-24364268 on Build GRCh38
rs768213443 Chr.X: 24364383-24364384 on Build GRCh38
rs747610071 Chr.X: 24364285-24364285 on Build GRCh38
rs750763370 Chr.X: 24364313-24364336 on Build GRCh38
rs745534241 Chr.X: 24364302-24364303 on Build GRCh38
rs374004998 Chr.X: 24364461-24364461 on Build GRCh38
rs769131314 Chr.X: 24364149-24364149 on Build GRCh38
rs760600694 Chr.X: 24364539-24364539 on Build GRCh38
rs748810658 Chr.X: 24364375-24364377 on Build GRCh38
rs867845567 Chr.X: 24364297-24364297 on Build GRCh38
rs752671213 Chr.X: 24364433-24364435 on Build GRCh38
rs775015453 Chr.X: 24364306-24364326 on Build GRCh38
rs866352747 Chr.X: 24364567-24364567 on Build GRCh38
rs866999802 Chr.X: 24364306-24364306 on Build GRCh38
rs774915478 Chr.X: 24364309-24364326 on Build GRCh38
rs766676433 Chr.X: 24364318-24364326 on Build GRCh38
rs764559977 Chr.X: 24364306-24364307 on Build GRCh38
rs370107923 Chr.X: 24364571-24364571 on Build GRCh38
rs775283877 Chr.X: 24364408-24364408 on Build GRCh38
rs754017985 Chr.X: 24364325-24364336 on Build GRCh38
rs377199102 Chr.X: 24364498-24364498 on Build GRCh38
rs763844539 Chr.X: 24364451-24364451 on Build GRCh38
rs747441497 Chr.X: 24364147-24364147 on Build GRCh38
rs769480317 Chr.X: 24364373-24364374 on Build GRCh38
rs35206911 Chr.X: 24364311-24364312 on Build GRCh38
rs750675412 Chr.X: 24364254-24364256 on Build GRCh38
rs374628239 Chr.X: 24364150-24364150 on Build GRCh38
rs767933568 Chr.X: 24364312-24364326 on Build GRCh38
rs754107569 Chr.X: 24364423-24364434 on Build GRCh38
rs750525009 Chr.X: 24364384-24364384 on Build GRCh38
rs778282259 Chr.X: 24364482-24364482 on Build GRCh38
rs769392191 Chr.X: 24364304-24364318 on Build GRCh38
rs745626175 Chr.X: 24364373-24364384 on Build GRCh38
rs769594874 Chr.X: 24364360-24364360 on Build GRCh38
rs779936825 Chr.X: 24364407-24364407 on Build GRCh38
rs772882046 Chr.X: 24364167-24364167 on Build GRCh38
rs2525863 Chr.X: 24364280-24364280 on Build GRCh38
rs915344197 Chr.X: 24364430-24364430 on Build GRCh38
rs780866415 Chr.X: 24364548-24364548 on Build GRCh38
rs879054015 Chr.X: 24364266-24364268 on Build GRCh38
rs185449787 Chr.X: 24364300-24364300 on Build GRCh38
rs746972712 Chr.X: 24364350-24364350 on Build GRCh38
rs779993614 Chr.X: 24364276-24364276 on Build GRCh38
rs766495756 Chr.X: 24364453-24364453 on Build GRCh38
rs776185252 Chr.X: 24364326-24364326 on Build GRCh38
rs776223470 Chr.X: 24364300-24364301 on Build GRCh38
rs756344204 Chr.X: 24364256-24364257 on Build GRCh38
rs759058006 Chr.X: 24364257-24364259 on Build GRCh38
rs755161340 Chr.X: 24364576-24364576 on Build GRCh38
rs866189148 Chr.X: 24364374-24364374 on Build GRCh38
rs935856190 Chr.X: 24364385-24364385 on Build GRCh38
rs751527991 Chr.X: 24364458-24364458 on Build GRCh38
rs770679887 Chr.X: 24364288-24364288 on Build GRCh38
rs773751492 Chr.X: 24364395-24364396 on Build GRCh38
rs760076453 Chr.X: 24364322-24364322 on Build GRCh38
rs370418088 Chr.X: 24364527-24364527 on Build GRCh38
rs755071805 Chr.X: 24364325-24364327 on Build GRCh38
rs759610958 Chr.X: 24364424-24364432 on Build GRCh38
rs754077958 Chr.X: 24364377-24364377 on Build GRCh38
rs2695489 Chr.X: 24364255-24364255 on Build GRCh38
rs757622357 Chr.X: 24364241-24364270 on Build GRCh38
rs756436220 Chr.X: 24364313-24364360 on Build GRCh38
rs772724573 Chr.X: 24364325-24364325 on Build GRCh38
rs762401096 Chr.X: 24364384-24364385 on Build GRCh38
rs758524685 Chr.X: 24364390-24364390 on Build GRCh38
rs777640278 Chr.X: 24364347-24364348 on Build GRCh38
rs767354652 Chr.X: 24364232-24364232 on Build GRCh38
rs112697166 Chr.X: 24364336-24364336 on Build GRCh38
rs765787789 Chr.X: 24364175-24364175 on Build GRCh38
rs781694819 Chr.X: 24364321-24364321 on Build GRCh38
rs770528188 Chr.X: 24364397-24364397 on Build GRCh38
rs769506288 Chr.X: 24364152-24364152 on Build GRCh38
rs773940697 Chr.X: 24364514-24364514 on Build GRCh38
rs771547146 Chr.X: 24364521-24364521 on Build GRCh38
rs377725511 Chr.X: 24364189-24364189 on Build GRCh38
rs774559877 Chr.X: 24364259-24364260 on Build GRCh38
rs748645908 Chr.X: 24364463-24364463 on Build GRCh38
rs770757321 Chr.X: 24364296-24364336 on Build GRCh38
rs779057260 Chr.X: 24364259-24364259 on Build GRCh38
rs762156994 Chr.X: 24364310-24364312 on Build GRCh38
rs199682391 Chr.X: 24364294-24364294 on Build GRCh38
rs766764063 Chr.X: 24364422-24364423 on Build GRCh38
rs765373877 Chr.X: 24364433-24364447 on Build GRCh38
rs924414740 Chr.X: 24364329-24364329 on Build GRCh38
rs759520099 Chr.X: 24364402-24364402 on Build GRCh38
rs867120724 Chr.X: 24364378-24364378 on Build GRCh38
rs752420776 Chr.X: 24364234-24364234 on Build GRCh38
rs866488895 Chr.X: 24364421-24364421 on Build GRCh38
rs758545001 Chr.X: 24364273-24364273 on Build GRCh38

View Full Product Details