Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27746719-27747219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L2
Entrez Gene ID
347442
Gene Name
DDB1 and CUL4 associated factor 8 like 2
Gene Aliases
WDR42C
UniGene
Hs.720885
Gene Chromosome Location
Chr.X: 27746809-27748821 on Build GRCh38
Transcripts
NM_001136533.1

SNP Detail

SNP ID Chromosome Location
rs757832106 Chr.X: 27747180-27747180 on Build GRCh38
rs780547381 Chr.X: 27746764-27746764 on Build GRCh38
rs376826675 Chr.X: 27747024-27747024 on Build GRCh38
rs776454813 Chr.X: 27746862-27746862 on Build GRCh38
rs1003128073 Chr.X: 27746968-27746968 on Build GRCh38
rs767437527 Chr.X: 27747145-27747145 on Build GRCh38
rs764963155 Chr.X: 27746879-27746879 on Build GRCh38
rs756619299 Chr.X: 27746923-27746923 on Build GRCh38
rs941334021 Chr.X: 27746998-27746998 on Build GRCh38
rs774569813 Chr.X: 27746888-27746888 on Build GRCh38
rs1040639326 Chr.X: 27746817-27746817 on Build GRCh38
rs752798391 Chr.X: 27746907-27746907 on Build GRCh38
rs768015516 Chr.X: 27746988-27746988 on Build GRCh38
rs748021210 Chr.X: 27747048-27747048 on Build GRCh38
rs768618306 Chr.X: 27746855-27746855 on Build GRCh38
rs772690346 Chr.X: 27747115-27747115 on Build GRCh38
rs375379571 Chr.X: 27747010-27747010 on Build GRCh38
rs113826309 Chr.X: 27746983-27746983 on Build GRCh38
rs183301101 Chr.X: 27746948-27746948 on Build GRCh38
rs928567467 Chr.X: 27746766-27746766 on Build GRCh38
rs776940792 Chr.X: 27746833-27746833 on Build GRCh38
rs947108536 Chr.X: 27746804-27746804 on Build GRCh38
rs61739703 Chr.X: 27747126-27747126 on Build GRCh38
rs745571354 Chr.X: 27746962-27746962 on Build GRCh38
rs762251646 Chr.X: 27747037-27747042 on Build GRCh38
rs779481870 Chr.X: 27746985-27746985 on Build GRCh38
rs769583415 Chr.X: 27747050-27747050 on Build GRCh38
rs750011372 Chr.X: 27746785-27746785 on Build GRCh38
rs760812430 Chr.X: 27747131-27747131 on Build GRCh38
rs746501868 Chr.X: 27746987-27746987 on Build GRCh38
rs759410595 Chr.X: 27746790-27746790 on Build GRCh38
rs767550922 Chr.X: 27746904-27746904 on Build GRCh38
rs62623432 Chr.X: 27746955-27746955 on Build GRCh38
rs757302570 Chr.X: 27746755-27746755 on Build GRCh38
rs990691409 Chr.X: 27747087-27747087 on Build GRCh38
rs900724463 Chr.X: 27746836-27746836 on Build GRCh38
rs946762686 Chr.X: 27746752-27746752 on Build GRCh38
rs199614366 Chr.X: 27747088-27747088 on Build GRCh38
rs1042438533 Chr.X: 27746783-27746783 on Build GRCh38
rs914407155 Chr.X: 27746802-27746802 on Build GRCh38
rs754344752 Chr.X: 27746945-27746945 on Build GRCh38
rs759738501 Chr.X: 27746903-27746903 on Build GRCh38
rs959528202 Chr.X: 27747060-27747060 on Build GRCh38
rs796489879 Chr.X: 27746908-27746908 on Build GRCh38
rs192806500 Chr.X: 27746939-27746939 on Build GRCh38
rs758559050 Chr.X: 27747119-27747119 on Build GRCh38
rs192908633 Chr.X: 27747165-27747165 on Build GRCh38
rs755720853 Chr.X: 27746874-27746874 on Build GRCh38

View Full Product Details