Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 31071781-31072229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTHL17
Entrez Gene ID
53940
Gene Name
ferritin heavy chain like 17
Gene Aliases
CT38
UniGene
Hs.333125
Gene Chromosome Location
Chr.X: 31071241-31072053 on Build GRCh38
Transcripts
NM_031894.2

SNP Detail

SNP ID Chromosome Location
rs367688767 Chr.X: 31072004-31072004 on Build GRCh38
rs886780108 Chr.X: 31072112-31072112 on Build GRCh38
rs757463494 Chr.X: 31072005-31072005 on Build GRCh38
rs752465456 Chr.X: 31072036-31072036 on Build GRCh38
rs774769349 Chr.X: 31071899-31071899 on Build GRCh38
rs142837724 Chr.X: 31071896-31071896 on Build GRCh38
rs777245976 Chr.X: 31071804-31071804 on Build GRCh38
rs551562864 Chr.X: 31071823-31071823 on Build GRCh38
rs866378534 Chr.X: 31072167-31072167 on Build GRCh38
rs930737606 Chr.X: 31072155-31072155 on Build GRCh38
rs767876220 Chr.X: 31071855-31071855 on Build GRCh38
rs770613475 Chr.X: 31071824-31071824 on Build GRCh38
rs12845687 Chr.X: 31072130-31072130 on Build GRCh38
rs1021577336 Chr.X: 31071906-31071906 on Build GRCh38
rs145075121 Chr.X: 31071900-31071900 on Build GRCh38
rs438870 Chr.X: 31071813-31071813 on Build GRCh38
rs748357093 Chr.X: 31071925-31071925 on Build GRCh38
rs779874259 Chr.X: 31071915-31071915 on Build GRCh38
rs189619097 Chr.X: 31071999-31071999 on Build GRCh38
rs931536191 Chr.X: 31072048-31072048 on Build GRCh38
rs771095024 Chr.X: 31071934-31071934 on Build GRCh38
rs372030733 Chr.X: 31071964-31071964 on Build GRCh38
rs899242122 Chr.X: 31072080-31072080 on Build GRCh38
rs750871154 Chr.X: 31071952-31071952 on Build GRCh38
rs764752594 Chr.X: 31071987-31071987 on Build GRCh38
rs140427727 Chr.X: 31071811-31071811 on Build GRCh38
rs1041209697 Chr.X: 31072114-31072114 on Build GRCh38
rs867269019 Chr.X: 31072154-31072154 on Build GRCh38
rs112525239 Chr.X: 31071981-31071981 on Build GRCh38
rs763621950 Chr.X: 31071828-31071828 on Build GRCh38
rs761340395 Chr.X: 31071983-31071983 on Build GRCh38
rs187655870 Chr.X: 31071831-31071831 on Build GRCh38
rs774744538 Chr.X: 31071945-31071945 on Build GRCh38
rs766151769 Chr.X: 31071998-31071998 on Build GRCh38
rs749384542 Chr.X: 31071932-31071932 on Build GRCh38
rs750373885 Chr.X: 31071992-31071992 on Build GRCh38
rs778523893 Chr.X: 31072017-31072017 on Build GRCh38
rs375072828 Chr.X: 31071994-31071994 on Build GRCh38
rs767675212 Chr.X: 31071910-31071910 on Build GRCh38
rs776196229 Chr.X: 31071976-31071976 on Build GRCh38
rs748349892 Chr.X: 31071816-31071816 on Build GRCh38
rs867038922 Chr.X: 31072131-31072131 on Build GRCh38
rs138241353 Chr.X: 31071815-31071815 on Build GRCh38
rs943330628 Chr.X: 31072037-31072037 on Build GRCh38
rs753817162 Chr.X: 31071805-31071805 on Build GRCh38
rs989605954 Chr.X: 31071991-31071991 on Build GRCh38
rs867657139 Chr.X: 31072161-31072161 on Build GRCh38
rs746525597 Chr.X: 31072129-31072129 on Build GRCh38
rs866174102 Chr.X: 31072139-31072139 on Build GRCh38
rs757819948 Chr.X: 31072066-31072066 on Build GRCh38
rs868261176 Chr.X: 31072124-31072124 on Build GRCh38
rs756401411 Chr.X: 31071926-31071926 on Build GRCh38
rs778057150 Chr.X: 31071930-31071930 on Build GRCh38
rs770923173 Chr.X: 31072168-31072168 on Build GRCh38
rs760292130 Chr.X: 31071876-31071876 on Build GRCh38
rs777394280 Chr.X: 31072113-31072113 on Build GRCh38
rs958947124 Chr.X: 31072052-31072052 on Build GRCh38
rs754761747 Chr.X: 31072002-31072002 on Build GRCh38
rs1048130547 Chr.X: 31072054-31072054 on Build GRCh38
rs759849862 Chr.X: 31071845-31071845 on Build GRCh38
rs1016480895 Chr.X: 31072159-31072159 on Build GRCh38
rs945620014 Chr.X: 31072003-31072003 on Build GRCh38
rs200895998 Chr.X: 31071877-31071877 on Build GRCh38
rs768938267 Chr.X: 31071893-31071893 on Build GRCh38
rs753337169 Chr.X: 31071880-31071880 on Build GRCh38
rs182102107 Chr.X: 31072012-31072012 on Build GRCh38
rs757185333 Chr.X: 31071809-31071809 on Build GRCh38
rs774856217 Chr.X: 31071833-31071833 on Build GRCh38
rs867496913 Chr.X: 31072136-31072136 on Build GRCh38
rs775921789 Chr.X: 31071954-31071954 on Build GRCh38
rs745659539 Chr.X: 31071812-31071812 on Build GRCh38
rs758147339 Chr.X: 31071914-31071914 on Build GRCh38
rs761193710 Chr.X: 31071975-31071975 on Build GRCh38
rs977918598 Chr.X: 31071968-31071968 on Build GRCh38
rs759407328 Chr.X: 31071864-31071864 on Build GRCh38
rs900889264 Chr.X: 31072121-31072121 on Build GRCh38

View Full Product Details