Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37846225-37846737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
UniGene
Hs.446392
Gene Chromosome Location
Chr.X: 37838836-37847636 on Build GRCh38
Transcripts
NM_006520.2

SNP Detail

SNP ID Chromosome Location
rs374042272 Chr.X: 37846306-37846306 on Build GRCh38
rs35612238 Chr.X: 37846359-37846359 on Build GRCh38
rs930974194 Chr.X: 37846426-37846426 on Build GRCh38
rs565732935 Chr.X: 37846416-37846416 on Build GRCh38
rs913337261 Chr.X: 37846528-37846528 on Build GRCh38
rs757569933 Chr.X: 37846326-37846326 on Build GRCh38
rs980759630 Chr.X: 37846462-37846462 on Build GRCh38
rs942650475 Chr.X: 37846516-37846516 on Build GRCh38
rs893922013 Chr.X: 37846597-37846597 on Build GRCh38
rs191291620 Chr.X: 37846633-37846633 on Build GRCh38
rs891111281 Chr.X: 37846614-37846614 on Build GRCh38
rs183070462 Chr.X: 37846580-37846580 on Build GRCh38
rs889702061 Chr.X: 37846511-37846511 on Build GRCh38
rs898384690 Chr.X: 37846681-37846681 on Build GRCh38
rs57226575 Chr.X: 37846540-37846540 on Build GRCh38
rs972488613 Chr.X: 37846379-37846379 on Build GRCh38
rs74872302 Chr.X: 37846571-37846571 on Build GRCh38
rs1024097271 Chr.X: 37846639-37846639 on Build GRCh38
rs757952914 Chr.X: 37846314-37846314 on Build GRCh38
rs901020529 Chr.X: 37846545-37846545 on Build GRCh38
rs139261541 Chr.X: 37846391-37846391 on Build GRCh38
rs766163513 Chr.X: 37846619-37846619 on Build GRCh38
rs753095836 Chr.X: 37846364-37846364 on Build GRCh38
rs761103838 Chr.X: 37846412-37846412 on Build GRCh38
rs144291298 Chr.X: 37846648-37846648 on Build GRCh38
rs186078339 Chr.X: 37846581-37846581 on Build GRCh38
rs1048562765 Chr.X: 37846488-37846488 on Build GRCh38
rs758079237 Chr.X: 37846590-37846590 on Build GRCh38
rs562793976 Chr.X: 37846697-37846697 on Build GRCh38
rs867332631 Chr.X: 37846355-37846355 on Build GRCh38
rs1004213960 Chr.X: 37846688-37846688 on Build GRCh38
rs777183944 Chr.X: 37846568-37846568 on Build GRCh38
rs867384404 Chr.X: 37846368-37846368 on Build GRCh38
rs370848392 Chr.X: 37846305-37846305 on Build GRCh38
rs779916659 Chr.X: 37846363-37846363 on Build GRCh38
rs201856049 Chr.X: 37846289-37846289 on Build GRCh38
rs946075404 Chr.X: 37846572-37846572 on Build GRCh38
rs201428720 Chr.X: 37846571-37846572 on Build GRCh38

View Full Product Details