Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40057019-40057515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
UniGene
Hs.659681
Gene Chromosome Location
Chr.X: 40051246-40177390 on Build GRCh38
Transcripts
NM_001123383.1 NM_001123384.1 NM_001123385.1 NM_017745.5 XM_005272616.1 XM_005272618.3 XM_005272619.3 XM_005272620.3 XM_006724536.3 XM_011543929.2 XM_011543930.1 XM_011543931.2 XM_017029615.1 XM_017029616.1

SNP Detail

SNP ID Chromosome Location
rs776933021 Chr.X: 40057362-40057362 on Build GRCh38
rs745562627 Chr.X: 40057344-40057344 on Build GRCh38
rs957731700 Chr.X: 40057463-40057463 on Build GRCh38
rs138741225 Chr.X: 40057262-40057262 on Build GRCh38
rs375830184 Chr.X: 40057190-40057190 on Build GRCh38
rs765759611 Chr.X: 40057200-40057200 on Build GRCh38
rs967726384 Chr.X: 40057442-40057442 on Build GRCh38
rs758631631 Chr.X: 40057169-40057169 on Build GRCh38
rs183866805 Chr.X: 40057383-40057383 on Build GRCh38
rs200240104 Chr.X: 40057105-40057105 on Build GRCh38
rs759580873 Chr.X: 40057046-40057046 on Build GRCh38
rs764631885 Chr.X: 40057420-40057420 on Build GRCh38
rs373177567 Chr.X: 40057490-40057490 on Build GRCh38
rs374206405 Chr.X: 40057172-40057172 on Build GRCh38
rs368796810 Chr.X: 40057217-40057217 on Build GRCh38
rs764494664 Chr.X: 40057189-40057189 on Build GRCh38
rs113039659 Chr.X: 40057431-40057431 on Build GRCh38
rs770555045 Chr.X: 40057146-40057146 on Build GRCh38
rs761093745 Chr.X: 40057187-40057187 on Build GRCh38
rs756063203 Chr.X: 40057316-40057316 on Build GRCh38
rs1024748459 Chr.X: 40057439-40057443 on Build GRCh38
rs781698751 Chr.X: 40057443-40057443 on Build GRCh38
rs190717058 Chr.X: 40057090-40057090 on Build GRCh38
rs762969297 Chr.X: 40057405-40057405 on Build GRCh38
rs971074076 Chr.X: 40057439-40057439 on Build GRCh38
rs766620661 Chr.X: 40057253-40057253 on Build GRCh38
rs372724488 Chr.X: 40057228-40057228 on Build GRCh38
rs375344311 Chr.X: 40057277-40057277 on Build GRCh38
rs748764772 Chr.X: 40057329-40057329 on Build GRCh38
rs763649282 Chr.X: 40057095-40057095 on Build GRCh38
rs914861368 Chr.X: 40057428-40057428 on Build GRCh38
rs921695453 Chr.X: 40057481-40057481 on Build GRCh38
rs199676230 Chr.X: 40057210-40057210 on Build GRCh38
rs750459436 Chr.X: 40057205-40057205 on Build GRCh38
rs781170619 Chr.X: 40057276-40057276 on Build GRCh38
rs923822114 Chr.X: 40057462-40057462 on Build GRCh38
rs536249514 Chr.X: 40057052-40057052 on Build GRCh38
rs369589363 Chr.X: 40057373-40057373 on Build GRCh38
rs748765914 Chr.X: 40057109-40057109 on Build GRCh38
rs895896638 Chr.X: 40057063-40057063 on Build GRCh38
rs989125556 Chr.X: 40057466-40057466 on Build GRCh38
rs770413249 Chr.X: 40057330-40057330 on Build GRCh38
rs760292402 Chr.X: 40057094-40057094 on Build GRCh38
rs754796420 Chr.X: 40057265-40057265 on Build GRCh38
rs28493631 Chr.X: 40057089-40057089 on Build GRCh38
rs778612759 Chr.X: 40057336-40057336 on Build GRCh38
rs771745244 Chr.X: 40057352-40057352 on Build GRCh38
rs767605588 Chr.X: 40057087-40057087 on Build GRCh38
rs777585092 Chr.X: 40057318-40057318 on Build GRCh38
rs755444417 Chr.X: 40057404-40057404 on Build GRCh38

View Full Product Details