Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40062657-40063161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
UniGene
Hs.659681
Gene Chromosome Location
Chr.X: 40051246-40177390 on Build GRCh38
Transcripts
NM_001123383.1 NM_001123384.1 NM_001123385.1 NM_017745.5 XM_005272616.1 XM_005272618.3 XM_005272619.3 XM_005272620.3 XM_006724536.3 XM_011543929.2 XM_011543930.1 XM_011543931.2 XM_017029615.1 XM_017029616.1

SNP Detail

SNP ID Chromosome Location
rs768842901 Chr.X: 40062865-40062865 on Build GRCh38
rs981265184 Chr.X: 40062913-40062913 on Build GRCh38
rs775698427 Chr.X: 40062753-40062753 on Build GRCh38
rs780712297 Chr.X: 40062856-40062856 on Build GRCh38
rs142866108 Chr.X: 40062905-40062905 on Build GRCh38
rs972573105 Chr.X: 40062968-40062968 on Build GRCh38
rs763853638 Chr.X: 40062938-40062938 on Build GRCh38
rs781229417 Chr.X: 40063118-40063120 on Build GRCh38
rs375650828 Chr.X: 40062795-40062795 on Build GRCh38
rs905710587 Chr.X: 40063038-40063038 on Build GRCh38
rs748201753 Chr.X: 40062992-40062992 on Build GRCh38
rs759722092 Chr.X: 40062696-40062696 on Build GRCh38
rs777973496 Chr.X: 40063012-40063012 on Build GRCh38
rs746425722 Chr.X: 40062725-40062725 on Build GRCh38
rs868100355 Chr.X: 40063011-40063011 on Build GRCh38
rs947103197 Chr.X: 40062789-40062789 on Build GRCh38
rs772939285 Chr.X: 40062896-40062896 on Build GRCh38
rs767103127 Chr.X: 40062915-40062915 on Build GRCh38
rs771933828 Chr.X: 40063054-40063054 on Build GRCh38
rs770038057 Chr.X: 40062878-40062878 on Build GRCh38
rs768694461 Chr.X: 40062752-40062752 on Build GRCh38
rs908375616 Chr.X: 40062941-40062941 on Build GRCh38
rs755466715 Chr.X: 40063115-40063115 on Build GRCh38
rs781252900 Chr.X: 40062866-40062866 on Build GRCh38
rs373762694 Chr.X: 40062911-40062911 on Build GRCh38
rs371205644 Chr.X: 40063047-40063047 on Build GRCh38
rs748445735 Chr.X: 40062868-40062868 on Build GRCh38
rs888552486 Chr.X: 40062985-40062985 on Build GRCh38
rs747127803 Chr.X: 40062740-40062740 on Build GRCh38
rs750973796 Chr.X: 40062767-40062767 on Build GRCh38
rs774132405 Chr.X: 40063046-40063046 on Build GRCh38
rs767068752 Chr.X: 40062781-40062781 on Build GRCh38
rs372510528 Chr.X: 40062966-40062966 on Build GRCh38
rs3810694 Chr.X: 40063106-40063106 on Build GRCh38
rs751902996 Chr.X: 40063107-40063107 on Build GRCh38
rs779548577 Chr.X: 40062708-40062708 on Build GRCh38
rs770766590 Chr.X: 40063042-40063042 on Build GRCh38
rs774275618 Chr.X: 40062684-40062684 on Build GRCh38
rs773106597 Chr.X: 40062755-40062755 on Build GRCh38
rs398124314 Chr.X: 40062741-40062741 on Build GRCh38
rs980033487 Chr.X: 40062698-40062698 on Build GRCh38
rs747055096 Chr.X: 40062857-40062857 on Build GRCh38
rs187172641 Chr.X: 40062945-40062945 on Build GRCh38
rs749073971 Chr.X: 40063026-40063026 on Build GRCh38
rs1032649430 Chr.X: 40062712-40062712 on Build GRCh38
rs760362391 Chr.X: 40062934-40062934 on Build GRCh38
rs969985732 Chr.X: 40062680-40062680 on Build GRCh38
rs765267478 Chr.X: 40063081-40063081 on Build GRCh38
rs182401624 Chr.X: 40062845-40062845 on Build GRCh38
rs886042842 Chr.X: 40063035-40063036 on Build GRCh38
rs577082253 Chr.X: 40062766-40062766 on Build GRCh38
rs763899537 Chr.X: 40062770-40062770 on Build GRCh38
rs863224850 Chr.X: 40062808-40062808 on Build GRCh38
rs758919208 Chr.X: 40062847-40062847 on Build GRCh38
rs752451175 Chr.X: 40062787-40062787 on Build GRCh38
rs370685925 Chr.X: 40062910-40062910 on Build GRCh38
rs775007855 Chr.X: 40063079-40063079 on Build GRCh38
rs930630046 Chr.X: 40062982-40062982 on Build GRCh38
rs193148630 Chr.X: 40063123-40063123 on Build GRCh38
rs199795707 Chr.X: 40063055-40063055 on Build GRCh38
rs772245711 Chr.X: 40062730-40062730 on Build GRCh38
rs374549542 Chr.X: 40063092-40063092 on Build GRCh38
rs781293725 Chr.X: 40062986-40062986 on Build GRCh38
rs752258567 Chr.X: 40062932-40062932 on Build GRCh38
rs752160903 Chr.X: 40062971-40062971 on Build GRCh38
rs758900726 Chr.X: 40062963-40062963 on Build GRCh38
rs757538161 Chr.X: 40062794-40062794 on Build GRCh38
rs760406187 Chr.X: 40062754-40062754 on Build GRCh38
rs750841214 Chr.X: 40062824-40062824 on Build GRCh38
rs754985350 Chr.X: 40062978-40062978 on Build GRCh38
rs776485818 Chr.X: 40063099-40063099 on Build GRCh38
rs960199081 Chr.X: 40063131-40063131 on Build GRCh38
rs779785937 Chr.X: 40063130-40063130 on Build GRCh38
rs763391206 Chr.X: 40063103-40063103 on Build GRCh38
rs775665144 Chr.X: 40062731-40062731 on Build GRCh38
rs772183536 Chr.X: 40062697-40062697 on Build GRCh38
rs376614903 Chr.X: 40062861-40062861 on Build GRCh38
rs769942209 Chr.X: 40063000-40063000 on Build GRCh38
rs780412607 Chr.X: 40062969-40062969 on Build GRCh38
rs770784599 Chr.X: 40062906-40062906 on Build GRCh38

View Full Product Details