Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40580864-40581356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP2
Entrez Gene ID
10159
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
UniGene
Hs.495960
Gene Chromosome Location
Chr.X: 40580964-40606637 on Build GRCh38
Transcripts
NM_005765.2

SNP Detail

SNP ID Chromosome Location
rs186401295 Chr.X: 40580946-40580946 on Build GRCh38
rs767723601 Chr.X: 40580918-40580918 on Build GRCh38
rs779526345 Chr.X: 40580962-40580962 on Build GRCh38
rs998661117 Chr.X: 40581328-40581328 on Build GRCh38
rs1028938617 Chr.X: 40580884-40580884 on Build GRCh38
rs765852654 Chr.X: 40581084-40581084 on Build GRCh38
rs770158244 Chr.X: 40581271-40581271 on Build GRCh38
rs964889973 Chr.X: 40581241-40581241 on Build GRCh38
rs771416167 Chr.X: 40581304-40581304 on Build GRCh38
rs183187874 Chr.X: 40580909-40580909 on Build GRCh38
rs867497089 Chr.X: 40581242-40581242 on Build GRCh38
rs1050677613 Chr.X: 40581220-40581220 on Build GRCh38
rs1009395841 Chr.X: 40580919-40580919 on Build GRCh38
rs761760848 Chr.X: 40581026-40581026 on Build GRCh38
rs889109771 Chr.X: 40581221-40581221 on Build GRCh38
rs772155205 Chr.X: 40580995-40580995 on Build GRCh38
rs771914631 Chr.X: 40581223-40581223 on Build GRCh38
rs963398976 Chr.X: 40581312-40581312 on Build GRCh38
rs752440514 Chr.X: 40580943-40580943 on Build GRCh38
rs954920397 Chr.X: 40581285-40581285 on Build GRCh38
rs377597654 Chr.X: 40581020-40581020 on Build GRCh38
rs751032860 Chr.X: 40581142-40581142 on Build GRCh38
rs11553343 Chr.X: 40580981-40580981 on Build GRCh38
rs760173936 Chr.X: 40580945-40580946 on Build GRCh38
rs930879993 Chr.X: 40581191-40581191 on Build GRCh38
rs1013257606 Chr.X: 40581188-40581188 on Build GRCh38
rs750334399 Chr.X: 40581074-40581074 on Build GRCh38
rs1042405959 Chr.X: 40581211-40581211 on Build GRCh38
rs748487635 Chr.X: 40581236-40581236 on Build GRCh38
rs903286696 Chr.X: 40581303-40581303 on Build GRCh38
rs991432643 Chr.X: 40580970-40580970 on Build GRCh38
rs111632930 Chr.X: 40581031-40581031 on Build GRCh38
rs11553345 Chr.X: 40581061-40581061 on Build GRCh38
rs939910278 Chr.X: 40580952-40580952 on Build GRCh38
rs779783772 Chr.X: 40581224-40581224 on Build GRCh38
rs977257090 Chr.X: 40580947-40580947 on Build GRCh38
rs754634267 Chr.X: 40580921-40580921 on Build GRCh38
rs937808566 Chr.X: 40581121-40581121 on Build GRCh38
rs957286838 Chr.X: 40580964-40580964 on Build GRCh38
rs11553342 Chr.X: 40581071-40581071 on Build GRCh38
rs940814818 Chr.X: 40581069-40581069 on Build GRCh38
rs762819551 Chr.X: 40581080-40581080 on Build GRCh38
rs745908628 Chr.X: 40580975-40580975 on Build GRCh38
rs1032905931 Chr.X: 40580948-40580948 on Build GRCh38
rs773422892 Chr.X: 40581290-40581290 on Build GRCh38
rs11553347 Chr.X: 40581018-40581018 on Build GRCh38
rs776564367 Chr.X: 40581024-40581024 on Build GRCh38
rs754617742 Chr.X: 40581154-40581154 on Build GRCh38
rs190867561 Chr.X: 40581250-40581250 on Build GRCh38
rs760894305 Chr.X: 40581007-40581007 on Build GRCh38
rs974009692 Chr.X: 40581318-40581319 on Build GRCh38
rs763567992 Chr.X: 40581298-40581298 on Build GRCh38
rs745354255 Chr.X: 40581169-40581169 on Build GRCh38
rs775458553 Chr.X: 40581006-40581006 on Build GRCh38

View Full Product Details