Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40628030-40628536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf38
Entrez Gene ID
159013
Gene Name
chromosome X open reading frame 38
Gene Aliases
-
UniGene
Hs.495961
Gene Chromosome Location
Chr.X: 40626921-40647678 on Build GRCh38
Transcripts
NM_144970.2 XM_005272589.2 XM_006724527.3 XM_006724528.2 XM_017029302.1 XM_017029303.1 XM_017029304.1 XM_017029305.1

SNP Detail

SNP ID Chromosome Location
rs964230425 Chr.X: 40628086-40628086 on Build GRCh38
rs939215718 Chr.X: 40628412-40628412 on Build GRCh38
rs2971598 Chr.X: 40628098-40628098 on Build GRCh38
rs926438008 Chr.X: 40628376-40628376 on Build GRCh38
rs1006481242 Chr.X: 40628387-40628387 on Build GRCh38
rs2968913 Chr.X: 40628504-40628504 on Build GRCh38
rs988552906 Chr.X: 40628165-40628165 on Build GRCh38
rs772176123 Chr.X: 40628075-40628075 on Build GRCh38
rs149420185 Chr.X: 40628488-40628488 on Build GRCh38
rs897774857 Chr.X: 40628485-40628485 on Build GRCh38
rs751479736 Chr.X: 40628296-40628296 on Build GRCh38
rs933058316 Chr.X: 40628334-40628334 on Build GRCh38
rs147728153 Chr.X: 40628088-40628088 on Build GRCh38
rs915292926 Chr.X: 40628223-40628223 on Build GRCh38
rs139353115 Chr.X: 40628426-40628426 on Build GRCh38
rs766347160 Chr.X: 40628175-40628175 on Build GRCh38
rs1037928103 Chr.X: 40628451-40628451 on Build GRCh38
rs906047895 Chr.X: 40628475-40628475 on Build GRCh38
rs984260216 Chr.X: 40628064-40628064 on Build GRCh38
rs1056301538 Chr.X: 40628474-40628474 on Build GRCh38
rs935345575 Chr.X: 40628104-40628104 on Build GRCh38
rs923054516 Chr.X: 40628237-40628237 on Build GRCh38
rs946812731 Chr.X: 40628327-40628327 on Build GRCh38
rs146709296 Chr.X: 40628354-40628354 on Build GRCh38
rs1047429818 Chr.X: 40628335-40628335 on Build GRCh38

View Full Product Details