Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41333291-41333790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
UniGene
Hs.728563
Gene Chromosome Location
Chr.X: 41333308-41364472 on Build GRCh38
Transcripts
NM_001356.4 NR_126093.1 XM_017029313.1 XM_017029314.1

SNP Detail

SNP ID Chromosome Location
rs775657273 Chr.X: 41333664-41333664 on Build GRCh38
rs772266060 Chr.X: 41333653-41333655 on Build GRCh38
rs779601270 Chr.X: 41333399-41333399 on Build GRCh38
rs760844928 Chr.X: 41333683-41333683 on Build GRCh38
rs934982593 Chr.X: 41333310-41333310 on Build GRCh38
rs12008629 Chr.X: 41333722-41333722 on Build GRCh38
rs17146019 Chr.X: 41333756-41333756 on Build GRCh38
rs371384625 Chr.X: 41333712-41333712 on Build GRCh38
rs759100418 Chr.X: 41333445-41333445 on Build GRCh38
rs1052431937 Chr.X: 41333364-41333364 on Build GRCh38
rs186664873 Chr.X: 41333724-41333724 on Build GRCh38
rs775054156 Chr.X: 41333615-41333615 on Build GRCh38
rs12008523 Chr.X: 41333436-41333436 on Build GRCh38
rs201279446 Chr.X: 41333711-41333712 on Build GRCh38
rs1026107327 Chr.X: 41333537-41333537 on Build GRCh38
rs1012274940 Chr.X: 41333326-41333326 on Build GRCh38
rs960142327 Chr.X: 41333481-41333481 on Build GRCh38
rs922337250 Chr.X: 41333633-41333633 on Build GRCh38
rs191196008 Chr.X: 41333714-41333714 on Build GRCh38
rs976581820 Chr.X: 41333628-41333628 on Build GRCh38
rs1052552553 Chr.X: 41333311-41333311 on Build GRCh38
rs994587130 Chr.X: 41333546-41333546 on Build GRCh38
rs963815515 Chr.X: 41333626-41333626 on Build GRCh38
rs770226900 Chr.X: 41333426-41333426 on Build GRCh38
rs12008630 Chr.X: 41333728-41333728 on Build GRCh38
rs189409694 Chr.X: 41333505-41333505 on Build GRCh38
rs750089833 Chr.X: 41333348-41333348 on Build GRCh38
rs892043666 Chr.X: 41333316-41333316 on Build GRCh38
rs771251274 Chr.X: 41333470-41333470 on Build GRCh38
rs984403493 Chr.X: 41333580-41333580 on Build GRCh38
rs180854597 Chr.X: 41333716-41333716 on Build GRCh38
rs145000030 Chr.X: 41333722-41333722 on Build GRCh38
rs186005081 Chr.X: 41333711-41333711 on Build GRCh38
rs750783092 Chr.X: 41333671-41333671 on Build GRCh38
rs894288176 Chr.X: 41333581-41333581 on Build GRCh38
rs935120664 Chr.X: 41333641-41333641 on Build GRCh38
rs946406890 Chr.X: 41333672-41333672 on Build GRCh38
rs905993768 Chr.X: 41333355-41333355 on Build GRCh38
rs943063422 Chr.X: 41333584-41333584 on Build GRCh38
rs748640548 Chr.X: 41333401-41333401 on Build GRCh38
rs958153192 Chr.X: 41333734-41333737 on Build GRCh38
rs762524723 Chr.X: 41333658-41333658 on Build GRCh38
rs1043323928 Chr.X: 41333335-41333335 on Build GRCh38
rs181738349 Chr.X: 41333541-41333541 on Build GRCh38
rs890141093 Chr.X: 41333383-41333383 on Build GRCh38
rs1034211779 Chr.X: 41333567-41333567 on Build GRCh38
rs914856638 Chr.X: 41333666-41333666 on Build GRCh38
rs184221562 Chr.X: 41333461-41333461 on Build GRCh38
rs757892014 Chr.X: 41333352-41333352 on Build GRCh38

View Full Product Details