Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44148137-44148641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs397895239 Chr.X: 44148256-44148257 on Build GRCh38
rs751334977 Chr.X: 44148614-44148614 on Build GRCh38
rs991573549 Chr.X: 44148509-44148509 on Build GRCh38
rs914737475 Chr.X: 44148204-44148205 on Build GRCh38
rs149112986 Chr.X: 44148171-44148171 on Build GRCh38
rs541262353 Chr.X: 44148392-44148392 on Build GRCh38
rs760013902 Chr.X: 44148226-44148227 on Build GRCh38
rs899170572 Chr.X: 44148192-44148192 on Build GRCh38
rs922984484 Chr.X: 44148368-44148368 on Build GRCh38
rs887532739 Chr.X: 44148560-44148560 on Build GRCh38
rs987596931 Chr.X: 44148253-44148253 on Build GRCh38
rs916979748 Chr.X: 44148512-44148512 on Build GRCh38
rs979005629 Chr.X: 44148283-44148283 on Build GRCh38
rs867899230 Chr.X: 44148224-44148224 on Build GRCh38
rs926193727 Chr.X: 44148345-44148345 on Build GRCh38
rs200149696 Chr.X: 44148179-44148180 on Build GRCh38
rs936954513 Chr.X: 44148382-44148382 on Build GRCh38
rs967571859 Chr.X: 44148274-44148274 on Build GRCh38
rs949823505 Chr.X: 44148578-44148578 on Build GRCh38
rs111241512 Chr.X: 44148230-44148230 on Build GRCh38
rs368363656 Chr.X: 44148230-44148230 on Build GRCh38
rs34501076 Chr.X: 44148257-44148258 on Build GRCh38
rs944315324 Chr.X: 44148234-44148235 on Build GRCh38
rs868373408 Chr.X: 44148225-44148225 on Build GRCh38
rs775319468 Chr.X: 44148492-44148492 on Build GRCh38
rs189543264 Chr.X: 44148227-44148227 on Build GRCh38
rs1020858309 Chr.X: 44148257-44148257 on Build GRCh38
rs776176854 Chr.X: 44148175-44148175 on Build GRCh38
rs968839050 Chr.X: 44148226-44148229 on Build GRCh38
rs866966802 Chr.X: 44148226-44148226 on Build GRCh38
rs763841814 Chr.X: 44148611-44148611 on Build GRCh38
rs186129923 Chr.X: 44148615-44148615 on Build GRCh38
rs762896687 Chr.X: 44148527-44148529 on Build GRCh38
rs34889083 Chr.X: 44148230-44148231 on Build GRCh38
rs574215392 Chr.X: 44148271-44148271 on Build GRCh38
rs1040429296 Chr.X: 44148341-44148341 on Build GRCh38
rs996136787 Chr.X: 44148219-44148219 on Build GRCh38

View Full Product Details