Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44148792-44149296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs768439834 Chr.X: 44148871-44148871 on Build GRCh38
rs748533621 Chr.X: 44148908-44148908 on Build GRCh38
rs80108125 Chr.X: 44148980-44148980 on Build GRCh38
rs758866914 Chr.X: 44148816-44148816 on Build GRCh38
rs1008901180 Chr.X: 44149117-44149117 on Build GRCh38
rs1044190966 Chr.X: 44149228-44149228 on Build GRCh38
rs1018480717 Chr.X: 44148995-44148995 on Build GRCh38
rs941163974 Chr.X: 44149059-44149059 on Build GRCh38
rs1041787797 Chr.X: 44149187-44149187 on Build GRCh38
rs184167369 Chr.X: 44149223-44149223 on Build GRCh38
rs941816452 Chr.X: 44149226-44149226 on Build GRCh38
rs759518776 Chr.X: 44148915-44148915 on Build GRCh38
rs1051054832 Chr.X: 44149102-44149102 on Build GRCh38
rs6610893 Chr.X: 44149070-44149070 on Build GRCh38
rs762835658 Chr.X: 44149129-44149129 on Build GRCh38
rs1000339366 Chr.X: 44149209-44149209 on Build GRCh38
rs908303853 Chr.X: 44149046-44149046 on Build GRCh38
rs756270485 Chr.X: 44148945-44148945 on Build GRCh38
rs749093591 Chr.X: 44148818-44148818 on Build GRCh38
rs1448865 Chr.X: 44149116-44149116 on Build GRCh38
rs867085679 Chr.X: 44148835-44148835 on Build GRCh38
rs768630482 Chr.X: 44149158-44149158 on Build GRCh38
rs780826437 Chr.X: 44149082-44149082 on Build GRCh38
rs374637588 Chr.X: 44148831-44148831 on Build GRCh38
rs745481515 Chr.X: 44149114-44149114 on Build GRCh38
rs773667612 Chr.X: 44148882-44148882 on Build GRCh38
rs751978799 Chr.X: 44148850-44148850 on Build GRCh38
rs958959466 Chr.X: 44148933-44148933 on Build GRCh38
rs755480992 Chr.X: 44148862-44148862 on Build GRCh38
rs1033157520 Chr.X: 44149213-44149213 on Build GRCh38
rs775040445 Chr.X: 44149126-44149126 on Build GRCh38
rs772364799 Chr.X: 44149062-44149062 on Build GRCh38
rs761709825 Chr.X: 44149245-44149245 on Build GRCh38
rs757555484 Chr.X: 44148941-44148941 on Build GRCh38
rs982937617 Chr.X: 44149014-44149014 on Build GRCh38
rs977068772 Chr.X: 44149048-44149048 on Build GRCh38
rs766761247 Chr.X: 44148907-44148907 on Build GRCh38
rs771672756 Chr.X: 44148852-44148852 on Build GRCh38
rs867513393 Chr.X: 44148890-44148890 on Build GRCh38
rs370842574 Chr.X: 44148885-44148885 on Build GRCh38
rs903279800 Chr.X: 44149203-44149203 on Build GRCh38
rs983099172 Chr.X: 44149212-44149212 on Build GRCh38
rs34729789 Chr.X: 44148853-44148853 on Build GRCh38
rs867320603 Chr.X: 44148879-44148879 on Build GRCh38
rs916890082 Chr.X: 44148991-44148991 on Build GRCh38
rs778950139 Chr.X: 44148827-44148827 on Build GRCh38
rs767384597 Chr.X: 44149256-44149256 on Build GRCh38
rs779213632 Chr.X: 44148900-44148901 on Build GRCh38
rs932547806 Chr.X: 44149079-44149079 on Build GRCh38
rs921157081 Chr.X: 44149075-44149075 on Build GRCh38
rs764578894 Chr.X: 44148819-44148819 on Build GRCh38
rs3747352 Chr.X: 44149225-44149225 on Build GRCh38
rs971122445 Chr.X: 44149005-44149005 on Build GRCh38
rs965993134 Chr.X: 44149037-44149037 on Build GRCh38
rs745349172 Chr.X: 44148839-44148839 on Build GRCh38
rs767649572 Chr.X: 44148931-44148931 on Build GRCh38

View Full Product Details