Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44232394-44232891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs772087258 Chr.X: 44232536-44232536 on Build GRCh38
rs1007421817 Chr.X: 44232701-44232701 on Build GRCh38
rs756531437 Chr.X: 44232586-44232586 on Build GRCh38
rs748673890 Chr.X: 44232438-44232438 on Build GRCh38
rs774755259 Chr.X: 44232498-44232498 on Build GRCh38
rs764112165 Chr.X: 44232541-44232541 on Build GRCh38
rs367613569 Chr.X: 44232645-44232645 on Build GRCh38
rs752386152 Chr.X: 44232866-44232866 on Build GRCh38
rs763399150 Chr.X: 44232486-44232486 on Build GRCh38
rs913078635 Chr.X: 44232775-44232775 on Build GRCh38
rs190139811 Chr.X: 44232556-44232556 on Build GRCh38
rs778456108 Chr.X: 44232587-44232587 on Build GRCh38
rs764994539 Chr.X: 44232828-44232828 on Build GRCh38
rs757461566 Chr.X: 44232595-44232595 on Build GRCh38
rs759278030 Chr.X: 44232748-44232748 on Build GRCh38
rs370389045 Chr.X: 44232642-44232642 on Build GRCh38
rs772351468 Chr.X: 44232607-44232607 on Build GRCh38
rs753247560 Chr.X: 44232601-44232601 on Build GRCh38
rs192827775 Chr.X: 44232585-44232585 on Build GRCh38
rs974508196 Chr.X: 44232698-44232698 on Build GRCh38
rs761348844 Chr.X: 44232562-44232562 on Build GRCh38
rs983123292 Chr.X: 44232799-44232799 on Build GRCh38
rs764799296 Chr.X: 44232655-44232655 on Build GRCh38
rs755368618 Chr.X: 44232573-44232573 on Build GRCh38
rs185264618 Chr.X: 44232437-44232437 on Build GRCh38
rs933077581 Chr.X: 44232744-44232744 on Build GRCh38
rs766269043 Chr.X: 44232604-44232604 on Build GRCh38
rs891696446 Chr.X: 44232451-44232451 on Build GRCh38
rs143101094 Chr.X: 44232644-44232644 on Build GRCh38
rs945977542 Chr.X: 44232786-44232786 on Build GRCh38
rs777944095 Chr.X: 44232465-44232465 on Build GRCh38
rs779273390 Chr.X: 44232599-44232599 on Build GRCh38
rs374146918 Chr.X: 44232706-44232706 on Build GRCh38
rs746849640 Chr.X: 44232643-44232643 on Build GRCh38
rs965499474 Chr.X: 44232729-44232729 on Build GRCh38
rs772786586 Chr.X: 44232688-44232688 on Build GRCh38
rs936638462 Chr.X: 44232837-44232837 on Build GRCh38
rs903836266 Chr.X: 44232835-44232835 on Build GRCh38
rs918816279 Chr.X: 44232764-44232764 on Build GRCh38
rs147496249 Chr.X: 44232713-44232713 on Build GRCh38
rs951629915 Chr.X: 44232779-44232779 on Build GRCh38
rs748361696 Chr.X: 44232449-44232449 on Build GRCh38
rs749348063 Chr.X: 44232592-44232592 on Build GRCh38
rs371172012 Chr.X: 44232563-44232563 on Build GRCh38
rs771239998 Chr.X: 44232491-44232491 on Build GRCh38
rs193126389 Chr.X: 44232436-44232436 on Build GRCh38
rs760469358 Chr.X: 44232537-44232537 on Build GRCh38
rs747994461 Chr.X: 44232853-44232853 on Build GRCh38
rs762459329 Chr.X: 44232704-44232704 on Build GRCh38
rs376768756 Chr.X: 44232527-44232527 on Build GRCh38
rs749666330 Chr.X: 44232547-44232547 on Build GRCh38
rs3747354 Chr.X: 44232582-44232582 on Build GRCh38

View Full Product Details