Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44523695-44524199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUNDC1
Entrez Gene ID
139341
Gene Name
FUN14 domain containing 1
Gene Aliases
-
UniGene
Hs.7549
Gene Chromosome Location
Chr.X: 44523639-44542975 on Build GRCh38
Transcripts
NM_173794.3

SNP Detail

SNP ID Chromosome Location
rs937402177 Chr.X: 44523857-44523857 on Build GRCh38
rs903605745 Chr.X: 44524121-44524121 on Build GRCh38
rs759799187 Chr.X: 44524143-44524143 on Build GRCh38
rs773647762 Chr.X: 44524163-44524163 on Build GRCh38
rs955284797 Chr.X: 44523719-44523719 on Build GRCh38
rs1017484958 Chr.X: 44524166-44524166 on Build GRCh38
rs1029536780 Chr.X: 44524012-44524012 on Build GRCh38
rs1043564011 Chr.X: 44523831-44523831 on Build GRCh38
rs946050676 Chr.X: 44523797-44523797 on Build GRCh38
rs1016980779 Chr.X: 44523990-44523990 on Build GRCh38
rs771018299 Chr.X: 44523859-44523859 on Build GRCh38
rs896182458 Chr.X: 44524062-44524062 on Build GRCh38
rs988489307 Chr.X: 44524066-44524066 on Build GRCh38
rs1005204980 Chr.X: 44523970-44523970 on Build GRCh38
rs929279958 Chr.X: 44523893-44523893 on Build GRCh38
rs866350783 Chr.X: 44523732-44523732 on Build GRCh38
rs188229974 Chr.X: 44523752-44523752 on Build GRCh38
rs772871328 Chr.X: 44523996-44523996 on Build GRCh38
rs963792330 Chr.X: 44523991-44523991 on Build GRCh38
rs913262405 Chr.X: 44523788-44523788 on Build GRCh38
rs1020597028 Chr.X: 44524108-44524108 on Build GRCh38
rs760427076 Chr.X: 44524088-44524088 on Build GRCh38
rs763018625 Chr.X: 44524167-44524167 on Build GRCh38
rs777654716 Chr.X: 44523758-44523758 on Build GRCh38
rs1057296063 Chr.X: 44524047-44524047 on Build GRCh38
rs747133127 Chr.X: 44524114-44524114 on Build GRCh38
rs886798255 Chr.X: 44523958-44523958 on Build GRCh38
rs1055890460 Chr.X: 44523894-44523894 on Build GRCh38
rs895951726 Chr.X: 44523897-44523897 on Build GRCh38
rs767413681 Chr.X: 44523982-44523982 on Build GRCh38
rs768811412 Chr.X: 44523849-44523849 on Build GRCh38
rs909306441 Chr.X: 44523901-44523901 on Build GRCh38
rs765977358 Chr.X: 44524155-44524155 on Build GRCh38
rs774446793 Chr.X: 44523907-44523907 on Build GRCh38
rs747133961 Chr.X: 44523790-44523790 on Build GRCh38
rs761777248 Chr.X: 44523933-44523933 on Build GRCh38
rs955189372 Chr.X: 44524025-44524025 on Build GRCh38
rs776803636 Chr.X: 44524038-44524038 on Build GRCh38
rs200619955 Chr.X: 44523746-44523746 on Build GRCh38
rs770934107 Chr.X: 44524169-44524169 on Build GRCh38

View Full Product Details