Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47218397-47218888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
UniGene
Hs.496068
Gene Chromosome Location
Chr.X: 47217881-47229997 on Build GRCh38
Transcripts
NM_006201.4 NM_033018.3 XM_011543921.1 XM_011543922.1 XM_011543923.1 XM_011543924.1 XM_011543925.2 XM_011543927.2 XM_011543928.1 XM_017029569.1 XM_017029570.1 XM_017029572.1 XM_017029573.1

SNP Detail

SNP ID Chromosome Location
rs782645257 Chr.X: 47218725-47218725 on Build GRCh38
rs979424044 Chr.X: 47218693-47218693 on Build GRCh38
rs372931400 Chr.X: 47218653-47218653 on Build GRCh38
rs900273973 Chr.X: 47218780-47218780 on Build GRCh38
rs200711551 Chr.X: 47218853-47218853 on Build GRCh38
rs781818134 Chr.X: 47218738-47218738 on Build GRCh38
rs992620091 Chr.X: 47218673-47218673 on Build GRCh38
rs925523989 Chr.X: 47218695-47218695 on Build GRCh38
rs781924240 Chr.X: 47218683-47218683 on Build GRCh38
rs891537124 Chr.X: 47218507-47218507 on Build GRCh38
rs782136869 Chr.X: 47218508-47218508 on Build GRCh38
rs904130371 Chr.X: 47218453-47218453 on Build GRCh38
rs782102458 Chr.X: 47218692-47218692 on Build GRCh38
rs782587244 Chr.X: 47218658-47218658 on Build GRCh38
rs782505433 Chr.X: 47218744-47218744 on Build GRCh38
rs900133635 Chr.X: 47218601-47218601 on Build GRCh38
rs782740034 Chr.X: 47218735-47218735 on Build GRCh38
rs782503069 Chr.X: 47218720-47218720 on Build GRCh38
rs782180619 Chr.X: 47218669-47218669 on Build GRCh38
rs782533683 Chr.X: 47218661-47218661 on Build GRCh38
rs782552877 Chr.X: 47218851-47218851 on Build GRCh38
rs374522627 Chr.X: 47218681-47218681 on Build GRCh38
rs891248741 Chr.X: 47218748-47218748 on Build GRCh38
rs782783778 Chr.X: 47218747-47218747 on Build GRCh38
rs781983314 Chr.X: 47218702-47218702 on Build GRCh38
rs1052548547 Chr.X: 47218722-47218722 on Build GRCh38
rs782081750 Chr.X: 47218635-47218635 on Build GRCh38
rs41305365 Chr.X: 47218704-47218704 on Build GRCh38
rs781824984 Chr.X: 47218505-47218505 on Build GRCh38
rs113804788 Chr.X: 47218736-47218736 on Build GRCh38
rs782696833 Chr.X: 47218671-47218671 on Build GRCh38
rs944227317 Chr.X: 47218753-47218753 on Build GRCh38
rs367687703 Chr.X: 47218617-47218617 on Build GRCh38
rs782342848 Chr.X: 47218697-47218697 on Build GRCh38
rs935480871 Chr.X: 47218488-47218488 on Build GRCh38
rs112889618 Chr.X: 47218805-47218805 on Build GRCh38
rs781908704 Chr.X: 47218750-47218750 on Build GRCh38
rs782047745 Chr.X: 47218731-47218731 on Build GRCh38
rs782159608 Chr.X: 47218710-47218710 on Build GRCh38
rs782535191 Chr.X: 47218643-47218643 on Build GRCh38
rs782762591 Chr.X: 47218525-47218525 on Build GRCh38
rs188385651 Chr.X: 47218659-47218659 on Build GRCh38
rs1009101666 Chr.X: 47218534-47218534 on Build GRCh38
rs781902655 Chr.X: 47218641-47218641 on Build GRCh38
rs782777311 Chr.X: 47218639-47218639 on Build GRCh38
rs996232419 Chr.X: 47218782-47218782 on Build GRCh38
rs1040498558 Chr.X: 47218759-47218759 on Build GRCh38
rs782696580 Chr.X: 47218662-47218662 on Build GRCh38
rs377475059 Chr.X: 47218774-47218774 on Build GRCh38
rs782784381 Chr.X: 47218717-47218717 on Build GRCh38
rs782598865 Chr.X: 47218680-47218680 on Build GRCh38
rs1018697711 Chr.X: 47218566-47218566 on Build GRCh38

View Full Product Details