Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47224532-47225030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
UniGene
Hs.496068
Gene Chromosome Location
Chr.X: 47217881-47229997 on Build GRCh38
Transcripts
NM_001170460.1 NM_006201.4 NM_033018.3 XM_011543920.2 XM_011543921.1 XM_011543922.1 XM_011543923.1 XM_011543924.1 XM_011543925.2 XM_011543926.1 XM_011543927.2 XM_011543928.1 XM_017029569.1 XM_017029570.1 XM_017029571.1 XM_017029572.1 XM_017029573.1

SNP Detail

SNP ID Chromosome Location
rs765036690 Chr.X: 47224898-47224898 on Build GRCh38
rs7887940 Chr.X: 47224970-47224970 on Build GRCh38
rs772088943 Chr.X: 47224571-47224571 on Build GRCh38
rs766415625 Chr.X: 47224915-47224915 on Build GRCh38
rs767105525 Chr.X: 47224939-47224939 on Build GRCh38
rs759641820 Chr.X: 47224560-47224560 on Build GRCh38
rs368594503 Chr.X: 47224976-47224976 on Build GRCh38
rs755698772 Chr.X: 47224966-47224966 on Build GRCh38
rs372290137 Chr.X: 47224572-47224572 on Build GRCh38
rs138055902 Chr.X: 47224750-47224750 on Build GRCh38
rs373980192 Chr.X: 47224896-47224896 on Build GRCh38
rs778247305 Chr.X: 47224795-47224797 on Build GRCh38
rs370897419 Chr.X: 47224836-47224836 on Build GRCh38
rs750350381 Chr.X: 47224677-47224677 on Build GRCh38
rs751082789 Chr.X: 47224762-47224762 on Build GRCh38
rs751174580 Chr.X: 47224923-47224923 on Build GRCh38
rs771284673 Chr.X: 47224772-47224772 on Build GRCh38
rs146600180 Chr.X: 47224633-47224633 on Build GRCh38
rs746422745 Chr.X: 47224780-47224780 on Build GRCh38
rs910301268 Chr.X: 47224796-47224796 on Build GRCh38
rs758590982 Chr.X: 47224775-47224777 on Build GRCh38
rs758007857 Chr.X: 47224680-47224680 on Build GRCh38
rs780885349 Chr.X: 47224763-47224763 on Build GRCh38
rs772533405 Chr.X: 47224785-47224785 on Build GRCh38
rs764369025 Chr.X: 47224581-47224581 on Build GRCh38
rs775608871 Chr.X: 47224798-47224798 on Build GRCh38
rs752344985 Chr.X: 47224949-47224949 on Build GRCh38
rs929869946 Chr.X: 47224640-47224640 on Build GRCh38
rs747076174 Chr.X: 47224800-47224800 on Build GRCh38
rs776848840 Chr.X: 47224887-47224887 on Build GRCh38
rs758959289 Chr.X: 47224977-47224977 on Build GRCh38
rs780478906 Chr.X: 47224981-47224981 on Build GRCh38
rs763025668 Chr.X: 47224911-47224911 on Build GRCh38
rs182623103 Chr.X: 47224938-47224938 on Build GRCh38
rs35558089 Chr.X: 47224589-47224590 on Build GRCh38
rs753630441 Chr.X: 47224597-47224597 on Build GRCh38
rs373143378 Chr.X: 47224576-47224576 on Build GRCh38
rs377055494 Chr.X: 47224910-47224910 on Build GRCh38
rs5952422 Chr.X: 47224638-47224638 on Build GRCh38
rs1012910960 Chr.X: 47224602-47224602 on Build GRCh38
rs923090906 Chr.X: 47224932-47224932 on Build GRCh38
rs749617674 Chr.X: 47224766-47224766 on Build GRCh38
rs768771708 Chr.X: 47224829-47224829 on Build GRCh38
rs750218615 Chr.X: 47224777-47224777 on Build GRCh38

View Full Product Details