Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47224752-47225262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
UniGene
Hs.496068
Gene Chromosome Location
Chr.X: 47217881-47229997 on Build GRCh38
Transcripts
NM_001170460.1 NM_006201.4 NM_033018.3 XM_011543920.2 XM_011543921.1 XM_011543922.1 XM_011543923.1 XM_011543924.1 XM_011543925.2 XM_011543926.1 XM_011543927.2 XM_011543928.1 XM_017029569.1 XM_017029570.1 XM_017029571.1 XM_017029572.1 XM_017029573.1

SNP Detail

SNP ID Chromosome Location
rs765036690 Chr.X: 47224898-47224898 on Build GRCh38
rs774389363 Chr.X: 47225147-47225147 on Build GRCh38
rs7887940 Chr.X: 47224970-47224970 on Build GRCh38
rs766415625 Chr.X: 47224915-47224915 on Build GRCh38
rs767105525 Chr.X: 47224939-47224939 on Build GRCh38
rs368594503 Chr.X: 47224976-47224976 on Build GRCh38
rs755698772 Chr.X: 47224966-47224966 on Build GRCh38
rs773209253 Chr.X: 47225131-47225131 on Build GRCh38
rs373980192 Chr.X: 47224896-47224896 on Build GRCh38
rs778247305 Chr.X: 47224795-47224797 on Build GRCh38
rs370897419 Chr.X: 47224836-47224836 on Build GRCh38
rs751174580 Chr.X: 47224923-47224923 on Build GRCh38
rs954881859 Chr.X: 47225161-47225161 on Build GRCh38
rs887723348 Chr.X: 47225135-47225135 on Build GRCh38
rs1021620979 Chr.X: 47225044-47225044 on Build GRCh38
rs747099075 Chr.X: 47225014-47225014 on Build GRCh38
rs768744808 Chr.X: 47225038-47225038 on Build GRCh38
rs776745143 Chr.X: 47225089-47225089 on Build GRCh38
rs746422745 Chr.X: 47224780-47224780 on Build GRCh38
rs747404909 Chr.X: 47225110-47225124 on Build GRCh38
rs910301268 Chr.X: 47224796-47224796 on Build GRCh38
rs758590982 Chr.X: 47224775-47224777 on Build GRCh38
rs772533405 Chr.X: 47224785-47224785 on Build GRCh38
rs375686085 Chr.X: 47225141-47225141 on Build GRCh38
rs1002257528 Chr.X: 47225168-47225168 on Build GRCh38
rs775608871 Chr.X: 47224798-47224798 on Build GRCh38
rs752344985 Chr.X: 47224949-47224949 on Build GRCh38
rs747076174 Chr.X: 47224800-47224800 on Build GRCh38
rs748455901 Chr.X: 47225111-47225111 on Build GRCh38
rs776848840 Chr.X: 47224887-47224887 on Build GRCh38
rs758959289 Chr.X: 47224977-47224977 on Build GRCh38
rs780478906 Chr.X: 47224981-47224981 on Build GRCh38
rs767103070 Chr.X: 47225153-47225153 on Build GRCh38
rs763025668 Chr.X: 47224911-47224911 on Build GRCh38
rs182623103 Chr.X: 47224938-47224938 on Build GRCh38
rs371115424 Chr.X: 47225124-47225124 on Build GRCh38
rs754645021 Chr.X: 47225192-47225192 on Build GRCh38
rs377055494 Chr.X: 47224910-47224910 on Build GRCh38
rs368252678 Chr.X: 47225152-47225152 on Build GRCh38
rs923090906 Chr.X: 47224932-47224932 on Build GRCh38
rs768771708 Chr.X: 47224829-47224829 on Build GRCh38
rs986325963 Chr.X: 47225225-47225225 on Build GRCh38
rs750218615 Chr.X: 47224777-47224777 on Build GRCh38
rs762830564 Chr.X: 47225136-47225136 on Build GRCh38

View Full Product Details