Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48476470-48476973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
UniGene
Hs.23170
Gene Chromosome Location
Chr.X: 48476021-48486364 on Build GRCh38
Transcripts
NM_001282157.1 NM_012280.3 NM_177439.2 XM_005272595.1 XM_011543893.2 XM_011543894.2 XM_011543895.1 XM_017029382.1

SNP Detail

SNP ID Chromosome Location
rs2268954 Chr.X: 48476947-48476947 on Build GRCh38
rs544522208 Chr.X: 48476907-48476907 on Build GRCh38
rs781872548 Chr.X: 48476943-48476943 on Build GRCh38
rs374407801 Chr.X: 48476676-48476676 on Build GRCh38
rs782487028 Chr.X: 48476856-48476856 on Build GRCh38
rs929513843 Chr.X: 48476660-48476660 on Build GRCh38
rs782601649 Chr.X: 48476902-48476902 on Build GRCh38
rs111495428 Chr.X: 48476852-48476852 on Build GRCh38
rs782708065 Chr.X: 48476740-48476740 on Build GRCh38
rs938114475 Chr.X: 48476711-48476711 on Build GRCh38
rs781839413 Chr.X: 48476772-48476772 on Build GRCh38
rs919522388 Chr.X: 48476646-48476646 on Build GRCh38
rs782147438 Chr.X: 48476684-48476684 on Build GRCh38
rs893848571 Chr.X: 48476816-48476816 on Build GRCh38
rs947243320 Chr.X: 48476836-48476836 on Build GRCh38
rs781925310 Chr.X: 48476610-48476610 on Build GRCh38
rs782725483 Chr.X: 48476729-48476729 on Build GRCh38
rs782069119 Chr.X: 48476673-48476673 on Build GRCh38
rs149307977 Chr.X: 48476850-48476850 on Build GRCh38

View Full Product Details