Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48477878-48478389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
UniGene
Hs.23170
Gene Chromosome Location
Chr.X: 48476021-48486364 on Build GRCh38
Transcripts
NM_001282157.1 NM_012280.3 NM_177439.2 XM_005272595.1 XM_011543893.2 XM_011543894.2 XM_011543895.1 XM_017029382.1

SNP Detail

SNP ID Chromosome Location
rs201218884 Chr.X: 48478194-48478194 on Build GRCh38
rs781820709 Chr.X: 48478164-48478164 on Build GRCh38
rs782527677 Chr.X: 48478177-48478177 on Build GRCh38
rs782566596 Chr.X: 48477997-48477997 on Build GRCh38
rs1051510439 Chr.X: 48477928-48477928 on Build GRCh38
rs782470986 Chr.X: 48478305-48478305 on Build GRCh38
rs782372624 Chr.X: 48478025-48478025 on Build GRCh38
rs782007645 Chr.X: 48478122-48478122 on Build GRCh38
rs782706835 Chr.X: 48478157-48478157 on Build GRCh38
rs782122614 Chr.X: 48478132-48478132 on Build GRCh38
rs782600419 Chr.X: 48478019-48478019 on Build GRCh38
rs782003032 Chr.X: 48478272-48478272 on Build GRCh38
rs781941978 Chr.X: 48477907-48477907 on Build GRCh38
rs782131203 Chr.X: 48478002-48478002 on Build GRCh38
rs943265337 Chr.X: 48477951-48477951 on Build GRCh38
rs782048148 Chr.X: 48477952-48477952 on Build GRCh38
rs781805078 Chr.X: 48478003-48478003 on Build GRCh38
rs782809967 Chr.X: 48478353-48478353 on Build GRCh38
rs781783918 Chr.X: 48478298-48478298 on Build GRCh38
rs200231120 Chr.X: 48478198-48478198 on Build GRCh38
rs943336972 Chr.X: 48478317-48478317 on Build GRCh38
rs782700340 Chr.X: 48478225-48478225 on Build GRCh38
rs782690548 Chr.X: 48478295-48478295 on Build GRCh38
rs140370809 Chr.X: 48477963-48477963 on Build GRCh38
rs978031080 Chr.X: 48478209-48478209 on Build GRCh38
rs781825374 Chr.X: 48477971-48477971 on Build GRCh38
rs782064472 Chr.X: 48478143-48478143 on Build GRCh38
rs911830385 Chr.X: 48477938-48477938 on Build GRCh38
rs782432954 Chr.X: 48478018-48478018 on Build GRCh38
rs782376827 Chr.X: 48478234-48478234 on Build GRCh38
rs797044864 Chr.X: 48478081-48478081 on Build GRCh38
rs781958574 Chr.X: 48477984-48477984 on Build GRCh38
rs968446623 Chr.X: 48478208-48478208 on Build GRCh38
rs782814222 Chr.X: 48478137-48478137 on Build GRCh38
rs782269307 Chr.X: 48478037-48478037 on Build GRCh38
rs782261840 Chr.X: 48478020-48478020 on Build GRCh38
rs1047519627 Chr.X: 48477986-48477986 on Build GRCh38
rs781889469 Chr.X: 48478141-48478141 on Build GRCh38
rs782657674 Chr.X: 48478360-48478360 on Build GRCh38
rs202148972 Chr.X: 48478059-48478059 on Build GRCh38
rs369359484 Chr.X: 48478001-48478001 on Build GRCh38
rs782678280 Chr.X: 48478035-48478035 on Build GRCh38
rs782351182 Chr.X: 48478082-48478082 on Build GRCh38
rs782318773 Chr.X: 48478039-48478039 on Build GRCh38
rs782105481 Chr.X: 48478078-48478078 on Build GRCh38
rs782171289 Chr.X: 48478219-48478219 on Build GRCh38

View Full Product Details