Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48481513-48482019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
UniGene
Hs.23170
Gene Chromosome Location
Chr.X: 48476021-48486364 on Build GRCh38
Transcripts
NM_001282157.1 NM_012280.3 NM_177439.2 XM_005272595.1 XM_011543893.2 XM_011543894.2 XM_011543895.1 XM_017029382.1

SNP Detail

SNP ID Chromosome Location
rs782705967 Chr.X: 48481666-48481666 on Build GRCh38
rs760394 Chr.X: 48481592-48481592 on Build GRCh38
rs781927192 Chr.X: 48481756-48481756 on Build GRCh38
rs782279701 Chr.X: 48481743-48481743 on Build GRCh38
rs782302473 Chr.X: 48481733-48481733 on Build GRCh38
rs1040742709 Chr.X: 48481816-48481816 on Build GRCh38
rs73483894 Chr.X: 48481624-48481624 on Build GRCh38
rs782778955 Chr.X: 48481639-48481639 on Build GRCh38
rs369650651 Chr.X: 48481598-48481598 on Build GRCh38
rs782544401 Chr.X: 48481641-48481641 on Build GRCh38
rs373042410 Chr.X: 48481600-48481600 on Build GRCh38
rs2070991 Chr.X: 48481597-48481597 on Build GRCh38
rs945276348 Chr.X: 48481815-48481815 on Build GRCh38
rs782388580 Chr.X: 48481901-48481901 on Build GRCh38
rs900825734 Chr.X: 48481921-48481921 on Build GRCh38
rs782236567 Chr.X: 48481586-48481586 on Build GRCh38
rs370432290 Chr.X: 48481667-48481667 on Build GRCh38
rs202218577 Chr.X: 48481758-48481758 on Build GRCh38
rs917226663 Chr.X: 48481587-48481587 on Build GRCh38
rs141084617 Chr.X: 48481714-48481714 on Build GRCh38
rs374511461 Chr.X: 48481755-48481755 on Build GRCh38
rs782795933 Chr.X: 48481843-48481843 on Build GRCh38
rs149550806 Chr.X: 48481640-48481640 on Build GRCh38
rs782228511 Chr.X: 48481556-48481556 on Build GRCh38
rs150266238 Chr.X: 48481944-48481944 on Build GRCh38
rs782200914 Chr.X: 48481763-48481763 on Build GRCh38
rs782444762 Chr.X: 48481535-48481535 on Build GRCh38
rs782597073 Chr.X: 48481681-48481681 on Build GRCh38
rs781887778 Chr.X: 48481932-48481932 on Build GRCh38
rs782684827 Chr.X: 48481734-48481734 on Build GRCh38
rs782345799 Chr.X: 48481762-48481762 on Build GRCh38
rs888170821 Chr.X: 48481984-48481984 on Build GRCh38
rs782716917 Chr.X: 48481597-48481598 on Build GRCh38
rs370068310 Chr.X: 48481576-48481576 on Build GRCh38
rs782483840 Chr.X: 48481575-48481575 on Build GRCh38
rs782787469 Chr.X: 48481816-48481816 on Build GRCh38

View Full Product Details