Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49064287-49064777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC120
Entrez Gene ID
90060
Gene Name
coiled-coil domain containing 120
Gene Aliases
JM11
UniGene
Hs.522643
Gene Chromosome Location
Chr.X: 49053432-49069858 on Build GRCh38
Transcripts
NM_001163321.2 NM_001163322.2 NM_001163323.2 NM_001271835.1 NM_001271836.1 NM_033626.3

SNP Detail

SNP ID Chromosome Location
rs782203688 Chr.X: 49064452-49064452 on Build GRCh38
rs782721807 Chr.X: 49064712-49064712 on Build GRCh38
rs781866928 Chr.X: 49064713-49064713 on Build GRCh38
rs782094614 Chr.X: 49064699-49064699 on Build GRCh38
rs927158132 Chr.X: 49064685-49064685 on Build GRCh38
rs782455534 Chr.X: 49064537-49064537 on Build GRCh38
rs1041965199 Chr.X: 49064476-49064476 on Build GRCh38
rs782350128 Chr.X: 49064330-49064330 on Build GRCh38
rs371924806 Chr.X: 49064602-49064602 on Build GRCh38
rs782551327 Chr.X: 49064425-49064425 on Build GRCh38
rs971303071 Chr.X: 49064656-49064656 on Build GRCh38
rs782192462 Chr.X: 49064430-49064430 on Build GRCh38
rs782040797 Chr.X: 49064472-49064472 on Build GRCh38
rs782485594 Chr.X: 49064421-49064421 on Build GRCh38
rs998116586 Chr.X: 49064510-49064510 on Build GRCh38
rs782611311 Chr.X: 49064638-49064638 on Build GRCh38
rs782004905 Chr.X: 49064485-49064485 on Build GRCh38
rs781943262 Chr.X: 49064342-49064342 on Build GRCh38
rs782683830 Chr.X: 49064328-49064328 on Build GRCh38
rs782816201 Chr.X: 49064394-49064394 on Build GRCh38
rs782697271 Chr.X: 49064538-49064538 on Build GRCh38
rs782636129 Chr.X: 49064564-49064564 on Build GRCh38
rs782146795 Chr.X: 49064648-49064648 on Build GRCh38
rs989935409 Chr.X: 49064733-49064733 on Build GRCh38
rs368629515 Chr.X: 49064594-49064594 on Build GRCh38
rs782028402 Chr.X: 49064644-49064644 on Build GRCh38
rs782127085 Chr.X: 49064352-49064352 on Build GRCh38
rs892808055 Chr.X: 49064470-49064470 on Build GRCh38
rs985235646 Chr.X: 49064315-49064315 on Build GRCh38
rs781853404 Chr.X: 49064423-49064423 on Build GRCh38
rs376279079 Chr.X: 49064643-49064643 on Build GRCh38
rs781838541 Chr.X: 49064420-49064420 on Build GRCh38
rs782426708 Chr.X: 49064437-49064437 on Build GRCh38
rs782329125 Chr.X: 49064649-49064649 on Build GRCh38
rs1029159808 Chr.X: 49064513-49064513 on Build GRCh38
rs782719151 Chr.X: 49064400-49064400 on Build GRCh38
rs782666031 Chr.X: 49064429-49064429 on Build GRCh38
rs782473362 Chr.X: 49064553-49064553 on Build GRCh38
rs781919463 Chr.X: 49064683-49064683 on Build GRCh38
rs374886808 Chr.X: 49064465-49064465 on Build GRCh38
rs371325682 Chr.X: 49064547-49064547 on Build GRCh38
rs781929013 Chr.X: 49064479-49064479 on Build GRCh38
rs782385920 Chr.X: 49064471-49064471 on Build GRCh38
rs782590983 Chr.X: 49064628-49064628 on Build GRCh38
rs914295140 Chr.X: 49064740-49064740 on Build GRCh38
rs372019456 Chr.X: 49064484-49064484 on Build GRCh38
rs374462954 Chr.X: 49064575-49064575 on Build GRCh38
rs782543796 Chr.X: 49064444-49064444 on Build GRCh38
rs782781153 Chr.X: 49064422-49064422 on Build GRCh38
rs782113608 Chr.X: 49064488-49064488 on Build GRCh38
rs782099877 Chr.X: 49064481-49064481 on Build GRCh38
rs781884117 Chr.X: 49064525-49064525 on Build GRCh38
rs782366460 Chr.X: 49064634-49064634 on Build GRCh38
rs782189699 Chr.X: 49064631-49064631 on Build GRCh38
rs200052201 Chr.X: 49064377-49064377 on Build GRCh38

View Full Product Details