Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49317188-49317690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAGE10
Entrez Gene ID
102724473
Gene Name
G antigen 10
Gene Aliases
GAGE-10
UniGene
Gene Chromosome Location
Chr.X: 49303646-49319844 on Build GRCh38
Transcripts
NM_001098413.3

SNP Detail

SNP ID Chromosome Location
rs199839509 Chr.X: 49317250-49317250 on Build GRCh38
rs781995439 Chr.X: 49317399-49317399 on Build GRCh38
rs782535908 Chr.X: 49317316-49317316 on Build GRCh38
rs373483037 Chr.X: 49317294-49317294 on Build GRCh38
rs782684716 Chr.X: 49317631-49317631 on Build GRCh38
rs192851367 Chr.X: 49317572-49317572 on Build GRCh38
rs782548476 Chr.X: 49317340-49317340 on Build GRCh38
rs781829043 Chr.X: 49317318-49317318 on Build GRCh38
rs781932937 Chr.X: 49317380-49317380 on Build GRCh38
rs369530119 Chr.X: 49317279-49317279 on Build GRCh38
rs782522632 Chr.X: 49317260-49317260 on Build GRCh38
rs782708184 Chr.X: 49317225-49317225 on Build GRCh38
rs782309902 Chr.X: 49317351-49317351 on Build GRCh38
rs782231380 Chr.X: 49317271-49317271 on Build GRCh38
rs782700498 Chr.X: 49317305-49317305 on Build GRCh38
rs782763482 Chr.X: 49317239-49317239 on Build GRCh38
rs782053450 Chr.X: 49317223-49317223 on Build GRCh38
rs782226804 Chr.X: 49317341-49317341 on Build GRCh38
rs782566770 Chr.X: 49317580-49317580 on Build GRCh38
rs781892850 Chr.X: 49317234-49317234 on Build GRCh38
rs782410822 Chr.X: 49317417-49317417 on Build GRCh38
rs782275724 Chr.X: 49317281-49317281 on Build GRCh38
rs782195009 Chr.X: 49317337-49317337 on Build GRCh38
rs782728749 Chr.X: 49317290-49317290 on Build GRCh38
rs782167554 Chr.X: 49317236-49317236 on Build GRCh38
rs191579278 Chr.X: 49317447-49317447 on Build GRCh38
rs781980927 Chr.X: 49317423-49317423 on Build GRCh38
rs782461131 Chr.X: 49317251-49317251 on Build GRCh38
rs782528877 Chr.X: 49317267-49317267 on Build GRCh38
rs781902104 Chr.X: 49317535-49317535 on Build GRCh38
rs782103819 Chr.X: 49317444-49317444 on Build GRCh38
rs201552194 Chr.X: 49317314-49317314 on Build GRCh38
rs782621638 Chr.X: 49317253-49317253 on Build GRCh38
rs781971300 Chr.X: 49317216-49317216 on Build GRCh38
rs149754081 Chr.X: 49317517-49317517 on Build GRCh38
rs781829730 Chr.X: 49317571-49317571 on Build GRCh38
rs782573695 Chr.X: 49317331-49317331 on Build GRCh38
rs186770618 Chr.X: 49317432-49317432 on Build GRCh38
rs2015487 Chr.X: 49317625-49317625 on Build GRCh38
rs782606725 Chr.X: 49317270-49317270 on Build GRCh38
rs781982798 Chr.X: 49317285-49317285 on Build GRCh38
rs782256324 Chr.X: 49317262-49317262 on Build GRCh38
rs5902444 Chr.X: 49317650-49317650 on Build GRCh38
rs782659658 Chr.X: 49317261-49317261 on Build GRCh38
rs782772490 Chr.X: 49317568-49317568 on Build GRCh38
rs139959382 Chr.X: 49317256-49317256 on Build GRCh38
rs782521684 Chr.X: 49317627-49317627 on Build GRCh38
rs781927908 Chr.X: 49317499-49317499 on Build GRCh38
rs376577624 Chr.X: 49317277-49317277 on Build GRCh38
rs782045267 Chr.X: 49317301-49317301 on Build GRCh38
rs782133013 Chr.X: 49317287-49317287 on Build GRCh38
rs782785305 Chr.X: 49317317-49317317 on Build GRCh38
rs782434290 Chr.X: 49317283-49317283 on Build GRCh38
rs369318514 Chr.X: 49317651-49317651 on Build GRCh38
rs188284351 Chr.X: 49317543-49317543 on Build GRCh38
rs373258005 Chr.X: 49317323-49317323 on Build GRCh38
rs368704784 Chr.X: 49317212-49317212 on Build GRCh38
rs375229404 Chr.X: 49317310-49317310 on Build GRCh38
rs782266665 Chr.X: 49317402-49317402 on Build GRCh38
rs182801059 Chr.X: 49317507-49317507 on Build GRCh38

View Full Product Details