Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103215490-103216019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEX4
Entrez Gene ID
56271
Gene Name
brain expressed X-linked 4
Gene Aliases
BEXL1
UniGene
Hs.184736
Gene Chromosome Location
Chr.X: 103215092-103217200 on Build GRCh38
Transcripts
NM_001080425.3 NM_001127688.2

SNP Detail

SNP ID Chromosome Location
rs769740690 Chr.X: 103215708-103215708 on Build GRCh38
rs931786600 Chr.X: 103215640-103215640 on Build GRCh38
rs764521238 Chr.X: 103215530-103215530 on Build GRCh38
rs946912445 Chr.X: 103215646-103215646 on Build GRCh38
rs987834612 Chr.X: 103215921-103215921 on Build GRCh38
rs1016852685 Chr.X: 103215853-103215853 on Build GRCh38
rs754363687 Chr.X: 103215565-103215570 on Build GRCh38
rs1035074180 Chr.X: 103215894-103215894 on Build GRCh38
rs749218752 Chr.X: 103215736-103215736 on Build GRCh38
rs1047872689 Chr.X: 103215812-103215812 on Build GRCh38
rs886923326 Chr.X: 103215815-103215815 on Build GRCh38
rs2273065 Chr.X: 103215886-103215886 on Build GRCh38
rs80101197 Chr.X: 103215611-103215611 on Build GRCh38
rs971811336 Chr.X: 103215887-103215887 on Build GRCh38
rs780333328 Chr.X: 103215683-103215683 on Build GRCh38
rs960695270 Chr.X: 103215622-103215622 on Build GRCh38
rs901862405 Chr.X: 103215714-103215714 on Build GRCh38
rs773238013 Chr.X: 103215721-103215721 on Build GRCh38
rs1040739082 Chr.X: 103215701-103215701 on Build GRCh38
rs1015849431 Chr.X: 103215557-103215557 on Build GRCh38
rs892418066 Chr.X: 103215838-103215838 on Build GRCh38
rs929353684 Chr.X: 103215780-103215780 on Build GRCh38
rs371171164 Chr.X: 103215737-103215737 on Build GRCh38
rs746027170 Chr.X: 103215644-103215644 on Build GRCh38
rs960415423 Chr.X: 103215900-103215900 on Build GRCh38
rs55914635 Chr.X: 103215625-103215625 on Build GRCh38
rs1005427643 Chr.X: 103215836-103215836 on Build GRCh38
rs11539751 Chr.X: 103215727-103215727 on Build GRCh38
rs867647231 Chr.X: 103215695-103215695 on Build GRCh38
rs149335987 Chr.X: 103215696-103215696 on Build GRCh38
rs943644310 Chr.X: 103215699-103215699 on Build GRCh38
rs901596416 Chr.X: 103215606-103215606 on Build GRCh38
rs914040757 Chr.X: 103215641-103215641 on Build GRCh38
rs980879041 Chr.X: 103215521-103215521 on Build GRCh38
rs758531968 Chr.X: 103215673-103215673 on Build GRCh38
rs1030895058 Chr.X: 103215777-103215777 on Build GRCh38
rs952216526 Chr.X: 103215947-103215947 on Build GRCh38
rs1034866366 Chr.X: 103215924-103215924 on Build GRCh38
rs968182322 Chr.X: 103215988-103215988 on Build GRCh38
rs192013004 Chr.X: 103215879-103215879 on Build GRCh38
rs988162727 Chr.X: 103215693-103215707 on Build GRCh38
rs181789076 Chr.X: 103215963-103215963 on Build GRCh38
rs985390905 Chr.X: 103215983-103215983 on Build GRCh38
rs979538284 Chr.X: 103215995-103215995 on Build GRCh38
rs867397351 Chr.X: 103215608-103215608 on Build GRCh38
rs774693014 Chr.X: 103215807-103215807 on Build GRCh38

View Full Product Details