Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103216842-103217344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEX4
Entrez Gene ID
56271
Gene Name
brain expressed X-linked 4
Gene Aliases
BEXL1
UniGene
Hs.184736
Gene Chromosome Location
Chr.X: 103215092-103217200 on Build GRCh38
Transcripts
NM_001080425.3 NM_001127688.2

SNP Detail

SNP ID Chromosome Location
rs41311733 Chr.X: 103216931-103216931 on Build GRCh38
rs1014966595 Chr.X: 103217154-103217155 on Build GRCh38
rs10543427 Chr.X: 103217068-103217069 on Build GRCh38
rs909183145 Chr.X: 103217271-103217271 on Build GRCh38
rs771073193 Chr.X: 103217174-103217174 on Build GRCh38
rs56828041 Chr.X: 103217146-103217147 on Build GRCh38
rs777887497 Chr.X: 103217017-103217017 on Build GRCh38
rs983721749 Chr.X: 103217199-103217199 on Build GRCh38
rs14661 Chr.X: 103217107-103217107 on Build GRCh38
rs765593007 Chr.X: 103217219-103217219 on Build GRCh38
rs985812771 Chr.X: 103217231-103217231 on Build GRCh38
rs143122720 Chr.X: 103217166-103217166 on Build GRCh38
rs950905781 Chr.X: 103217176-103217176 on Build GRCh38
rs369883322 Chr.X: 103217119-103217119 on Build GRCh38
rs913696665 Chr.X: 103216908-103216908 on Build GRCh38
rs745310123 Chr.X: 103216964-103216964 on Build GRCh38
rs925584218 Chr.X: 103217240-103217240 on Build GRCh38
rs1046317772 Chr.X: 103216976-103216976 on Build GRCh38
rs60244900 Chr.X: 103217296-103217296 on Build GRCh38
rs558834029 Chr.X: 103217134-103217135 on Build GRCh38
rs748344741 Chr.X: 103217220-103217223 on Build GRCh38
rs1052190714 Chr.X: 103216885-103216885 on Build GRCh38
rs543816507 Chr.X: 103217154-103217154 on Build GRCh38
rs751253183 Chr.X: 103217135-103217136 on Build GRCh38
rs953781895 Chr.X: 103217206-103217206 on Build GRCh38
rs150729695 Chr.X: 103217169-103217169 on Build GRCh38
rs1000558680 Chr.X: 103217020-103217020 on Build GRCh38
rs1030569380 Chr.X: 103217168-103217168 on Build GRCh38
rs962163663 Chr.X: 103217161-103217161 on Build GRCh38
rs1042014193 Chr.X: 103216981-103216981 on Build GRCh38
rs770760822 Chr.X: 103216929-103216929 on Build GRCh38
rs1006230329 Chr.X: 103217153-103217155 on Build GRCh38
rs202135929 Chr.X: 103217133-103217134 on Build GRCh38
rs905135216 Chr.X: 103217038-103217038 on Build GRCh38
rs747158280 Chr.X: 103217162-103217162 on Build GRCh38
rs771646179 Chr.X: 103217056-103217056 on Build GRCh38
rs1056236153 Chr.X: 103217118-103217118 on Build GRCh38
rs374766065 Chr.X: 103217147-103217147 on Build GRCh38
rs868735905 Chr.X: 103217064-103217065 on Build GRCh38
rs768363643 Chr.X: 103217145-103217145 on Build GRCh38

View Full Product Details