Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103707255-103707754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA4
Entrez Gene ID
441509
Gene Name
glycine receptor alpha 4
Gene Aliases
-
UniGene
Hs.533289
Gene Chromosome Location
Chr.X: 103707344-103728624 on Build GRCh38
Transcripts
NM_001024452.2

SNP Detail

SNP ID Chromosome Location
rs867975044 Chr.X: 103707426-103707426 on Build GRCh38
rs754855042 Chr.X: 103707505-103707505 on Build GRCh38
rs752078344 Chr.X: 103707548-103707548 on Build GRCh38
rs931439147 Chr.X: 103707714-103707714 on Build GRCh38
rs757652325 Chr.X: 103707461-103707461 on Build GRCh38
rs763039377 Chr.X: 103707535-103707535 on Build GRCh38
rs779890330 Chr.X: 103707463-103707463 on Build GRCh38
rs185557554 Chr.X: 103707365-103707365 on Build GRCh38
rs765432712 Chr.X: 103707506-103707506 on Build GRCh38
rs754022310 Chr.X: 103707423-103707423 on Build GRCh38
rs748923503 Chr.X: 103707464-103707464 on Build GRCh38
rs757911157 Chr.X: 103707621-103707621 on Build GRCh38
rs995477304 Chr.X: 103707403-103707403 on Build GRCh38
rs960300559 Chr.X: 103707562-103707562 on Build GRCh38
rs368872071 Chr.X: 103707392-103707392 on Build GRCh38
rs762577672 Chr.X: 103707720-103707721 on Build GRCh38
rs61744012 Chr.X: 103707415-103707415 on Build GRCh38
rs776888215 Chr.X: 103707497-103707497 on Build GRCh38
rs747308912 Chr.X: 103707364-103707364 on Build GRCh38
rs766611634 Chr.X: 103707418-103707418 on Build GRCh38
rs777996283 Chr.X: 103707363-103707363 on Build GRCh38
rs772967436 Chr.X: 103707356-103707356 on Build GRCh38
rs940185667 Chr.X: 103707645-103707645 on Build GRCh38
rs774567005 Chr.X: 103707347-103707347 on Build GRCh38
rs754966600 Chr.X: 103707319-103707319 on Build GRCh38
rs1037044379 Chr.X: 103707660-103707660 on Build GRCh38
rs200206348 Chr.X: 103707318-103707318 on Build GRCh38
rs779506083 Chr.X: 103707327-103707327 on Build GRCh38
rs180793883 Chr.X: 103707627-103707627 on Build GRCh38
rs992929757 Chr.X: 103707583-103707583 on Build GRCh38
rs374306209 Chr.X: 103707501-103707501 on Build GRCh38
rs954306988 Chr.X: 103707597-103707597 on Build GRCh38
rs145491863 Chr.X: 103707507-103707507 on Build GRCh38
rs746839773 Chr.X: 103707490-103707490 on Build GRCh38
rs750271684 Chr.X: 103707600-103707600 on Build GRCh38
rs774391126 Chr.X: 103707399-103707399 on Build GRCh38
rs149690029 Chr.X: 103707440-103707440 on Build GRCh38
rs372922558 Chr.X: 103707409-103707409 on Build GRCh38
rs771919736 Chr.X: 103707487-103707487 on Build GRCh38
rs778365732 Chr.X: 103707459-103707459 on Build GRCh38
rs928937484 Chr.X: 103707640-103707641 on Build GRCh38
rs764726337 Chr.X: 103707541-103707541 on Build GRCh38
rs758833866 Chr.X: 103707458-103707458 on Build GRCh38
rs568610725 Chr.X: 103707436-103707436 on Build GRCh38
rs973558458 Chr.X: 103707625-103707625 on Build GRCh38
rs761646273 Chr.X: 103707400-103707400 on Build GRCh38
rs753313587 Chr.X: 103707447-103707447 on Build GRCh38
rs189978029 Chr.X: 103707477-103707477 on Build GRCh38
rs911781782 Chr.X: 103707711-103707711 on Build GRCh38
rs747630275 Chr.X: 103707478-103707478 on Build GRCh38
rs748835841 Chr.X: 103707389-103707389 on Build GRCh38
rs770682435 Chr.X: 103707496-103707496 on Build GRCh38
rs1021747603 Chr.X: 103707483-103707483 on Build GRCh38
rs779800490 Chr.X: 103707369-103707369 on Build GRCh38
rs753763718 Chr.X: 103707411-103707411 on Build GRCh38
rs752681807 Chr.X: 103707642-103707642 on Build GRCh38
rs971654387 Chr.X: 103707452-103707452 on Build GRCh38
rs754570437 Chr.X: 103707471-103707471 on Build GRCh38
rs773086250 Chr.X: 103707488-103707488 on Build GRCh38
rs920006093 Chr.X: 103707706-103707706 on Build GRCh38
rs758854189 Chr.X: 103707353-103707353 on Build GRCh38
rs377077175 Chr.X: 103707352-103707352 on Build GRCh38
rs745785741 Chr.X: 103707355-103707356 on Build GRCh38

View Full Product Details