Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 104166594-104167105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM199X
Entrez Gene ID
139231
Gene Name
family with sequence similarity 199, X-linked
Gene Aliases
CXorf39
UniGene
Hs.496535
Gene Chromosome Location
Chr.X: 104157074-104195902 on Build GRCh38
Transcripts
NM_207318.3 XM_005262079.2 XM_017029270.1

SNP Detail

SNP ID Chromosome Location
rs782082661 Chr.X: 104166784-104166784 on Build GRCh38
rs782402739 Chr.X: 104166881-104166881 on Build GRCh38
rs782345485 Chr.X: 104166890-104166890 on Build GRCh38
rs782138189 Chr.X: 104167030-104167030 on Build GRCh38
rs782743789 Chr.X: 104166976-104166976 on Build GRCh38
rs894490132 Chr.X: 104166852-104166852 on Build GRCh38
rs782229637 Chr.X: 104166887-104166887 on Build GRCh38
rs782421549 Chr.X: 104166928-104166928 on Build GRCh38
rs782443781 Chr.X: 104166850-104166850 on Build GRCh38
rs782389866 Chr.X: 104166716-104166716 on Build GRCh38
rs781957075 Chr.X: 104166992-104166992 on Build GRCh38
rs782666525 Chr.X: 104166855-104166855 on Build GRCh38
rs782117470 Chr.X: 104166966-104166966 on Build GRCh38
rs782491429 Chr.X: 104166807-104166807 on Build GRCh38
rs782507584 Chr.X: 104166870-104166870 on Build GRCh38
rs1045759626 Chr.X: 104166932-104166932 on Build GRCh38
rs782531481 Chr.X: 104167029-104167030 on Build GRCh38
rs140881112 Chr.X: 104166803-104166803 on Build GRCh38
rs782148249 Chr.X: 104166802-104166802 on Build GRCh38
rs781901070 Chr.X: 104167028-104167028 on Build GRCh38
rs782034759 Chr.X: 104166798-104166798 on Build GRCh38
rs782299666 Chr.X: 104166925-104166925 on Build GRCh38
rs781981441 Chr.X: 104166745-104166745 on Build GRCh38
rs782693505 Chr.X: 104167018-104167018 on Build GRCh38
rs782261771 Chr.X: 104166737-104166737 on Build GRCh38
rs1031269743 Chr.X: 104167044-104167044 on Build GRCh38
rs781814096 Chr.X: 104166815-104166815 on Build GRCh38
rs781921538 Chr.X: 104166904-104166904 on Build GRCh38
rs781940154 Chr.X: 104167008-104167008 on Build GRCh38
rs372440871 Chr.X: 104166772-104166772 on Build GRCh38
rs781864420 Chr.X: 104166806-104166806 on Build GRCh38
rs1049138430 Chr.X: 104166818-104166818 on Build GRCh38
rs150154487 Chr.X: 104166859-104166859 on Build GRCh38
rs782075498 Chr.X: 104166995-104166995 on Build GRCh38
rs782211407 Chr.X: 104166698-104166698 on Build GRCh38

View Full Product Details