Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 104187956-104188461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM199X
Entrez Gene ID
139231
Gene Name
family with sequence similarity 199, X-linked
Gene Aliases
CXorf39
UniGene
Hs.496535
Gene Chromosome Location
Chr.X: 104157074-104195902 on Build GRCh38
Transcripts
NM_207318.3 XM_005262079.2 XM_017029270.1

SNP Detail

SNP ID Chromosome Location
rs782433717 Chr.X: 104188187-104188187 on Build GRCh38
rs1042313763 Chr.X: 104188341-104188341 on Build GRCh38
rs200335091 Chr.X: 104188199-104188199 on Build GRCh38
rs782037944 Chr.X: 104188056-104188056 on Build GRCh38
rs782257477 Chr.X: 104188234-104188234 on Build GRCh38
rs964781319 Chr.X: 104188417-104188417 on Build GRCh38
rs903709869 Chr.X: 104188397-104188397 on Build GRCh38
rs782304247 Chr.X: 104188304-104188305 on Build GRCh38
rs781803967 Chr.X: 104188138-104188138 on Build GRCh38
rs782515100 Chr.X: 104188235-104188235 on Build GRCh38
rs782211509 Chr.X: 104188083-104188083 on Build GRCh38
rs781925790 Chr.X: 104188336-104188336 on Build GRCh38
rs1009871599 Chr.X: 104188248-104188248 on Build GRCh38
rs782764049 Chr.X: 104188092-104188092 on Build GRCh38
rs782409299 Chr.X: 104188353-104188353 on Build GRCh38
rs782677312 Chr.X: 104188308-104188309 on Build GRCh38
rs782333951 Chr.X: 104188323-104188323 on Build GRCh38
rs896423622 Chr.X: 104188217-104188217 on Build GRCh38
rs782137078 Chr.X: 104188078-104188078 on Build GRCh38
rs782719161 Chr.X: 104188048-104188048 on Build GRCh38
rs781961664 Chr.X: 104188023-104188023 on Build GRCh38
rs782250300 Chr.X: 104187997-104187997 on Build GRCh38
rs938706251 Chr.X: 104188154-104188154 on Build GRCh38
rs782303925 Chr.X: 104188342-104188342 on Build GRCh38
rs782362898 Chr.X: 104188416-104188416 on Build GRCh38
rs782612392 Chr.X: 104188271-104188271 on Build GRCh38
rs782361092 Chr.X: 104188153-104188153 on Build GRCh38
rs782482950 Chr.X: 104188107-104188107 on Build GRCh38
rs782363704 Chr.X: 104188011-104188011 on Build GRCh38
rs782215701 Chr.X: 104188412-104188412 on Build GRCh38
rs964446062 Chr.X: 104187989-104187989 on Build GRCh38
rs923554384 Chr.X: 104187998-104187998 on Build GRCh38
rs782074075 Chr.X: 104188044-104188044 on Build GRCh38
rs782690336 Chr.X: 104188229-104188229 on Build GRCh38
rs782727044 Chr.X: 104188119-104188119 on Build GRCh38
rs782588676 Chr.X: 104188313-104188313 on Build GRCh38
rs781850251 Chr.X: 104188099-104188099 on Build GRCh38
rs782316945 Chr.X: 104188139-104188140 on Build GRCh38

View Full Product Details