Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 104189367-104189876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM199X
Entrez Gene ID
139231
Gene Name
family with sequence similarity 199, X-linked
Gene Aliases
CXorf39
UniGene
Hs.496535
Gene Chromosome Location
Chr.X: 104157074-104195902 on Build GRCh38
Transcripts
NM_207318.3 XM_005262079.2 XM_017029270.1

SNP Detail

SNP ID Chromosome Location
rs782268718 Chr.X: 104189696-104189696 on Build GRCh38
rs782702516 Chr.X: 104189648-104189648 on Build GRCh38
rs782612367 Chr.X: 104189798-104189798 on Build GRCh38
rs782273157 Chr.X: 104189807-104189807 on Build GRCh38
rs56179401 Chr.X: 104189479-104189479 on Build GRCh38
rs782753759 Chr.X: 104189681-104189681 on Build GRCh38
rs940418233 Chr.X: 104189758-104189758 on Build GRCh38
rs373647361 Chr.X: 104189679-104189679 on Build GRCh38
rs933157931 Chr.X: 104189708-104189708 on Build GRCh38
rs781953803 Chr.X: 104189624-104189624 on Build GRCh38
rs942663998 Chr.X: 104189423-104189423 on Build GRCh38
rs782658104 Chr.X: 104189797-104189797 on Build GRCh38
rs202121510 Chr.X: 104189680-104189680 on Build GRCh38
rs376219887 Chr.X: 104189643-104189643 on Build GRCh38
rs367786394 Chr.X: 104189821-104189821 on Build GRCh38
rs781786612 Chr.X: 104189753-104189753 on Build GRCh38
rs782071312 Chr.X: 104189625-104189625 on Build GRCh38
rs781994619 Chr.X: 104189563-104189563 on Build GRCh38
rs782547440 Chr.X: 104189796-104189796 on Build GRCh38
rs782461144 Chr.X: 104189712-104189712 on Build GRCh38
rs782352700 Chr.X: 104189587-104189587 on Build GRCh38
rs925914554 Chr.X: 104189407-104189407 on Build GRCh38
rs782104816 Chr.X: 104189585-104189585 on Build GRCh38
rs781836368 Chr.X: 104189691-104189691 on Build GRCh38
rs143955815 Chr.X: 104189404-104189404 on Build GRCh38
rs140348929 Chr.X: 104189718-104189718 on Build GRCh38
rs1037401945 Chr.X: 104189770-104189770 on Build GRCh38

View Full Product Details