Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106632621-106633164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf57
Entrez Gene ID
55086
Gene Name
chromosome X open reading frame 57
Gene Aliases
-
UniGene
Hs.274267
Gene Chromosome Location
Chr.X: 106611930-106679443 on Build GRCh38
Transcripts
NM_001184782.1 NM_018015.5

SNP Detail

SNP ID Chromosome Location
rs754469555 Chr.X: 106632767-106632767 on Build GRCh38
rs760344526 Chr.X: 106633030-106633030 on Build GRCh38
rs371791774 Chr.X: 106633067-106633067 on Build GRCh38
rs375031968 Chr.X: 106633088-106633088 on Build GRCh38
rs34578476 Chr.X: 106632865-106632866 on Build GRCh38
rs764438772 Chr.X: 106632698-106632698 on Build GRCh38
rs766199374 Chr.X: 106633047-106633047 on Build GRCh38
rs1346255 Chr.X: 106632644-106632644 on Build GRCh38
rs746381872 Chr.X: 106632949-106632949 on Build GRCh38
rs976528927 Chr.X: 106632758-106632758 on Build GRCh38
rs775626783 Chr.X: 106632669-106632669 on Build GRCh38
rs746708357 Chr.X: 106633094-106633094 on Build GRCh38
rs997361698 Chr.X: 106632813-106632813 on Build GRCh38
rs368813445 Chr.X: 106632888-106632888 on Build GRCh38
rs775953253 Chr.X: 106633022-106633022 on Build GRCh38
rs755982592 Chr.X: 106632736-106632736 on Build GRCh38
rs950118664 Chr.X: 106633075-106633075 on Build GRCh38
rs183727526 Chr.X: 106632742-106632742 on Build GRCh38
rs746293898 Chr.X: 106633114-106633114 on Build GRCh38
rs980142602 Chr.X: 106633025-106633025 on Build GRCh38
rs963114465 Chr.X: 106632980-106632980 on Build GRCh38
rs776021544 Chr.X: 106633126-106633126 on Build GRCh38
rs189339038 Chr.X: 106632851-106632851 on Build GRCh38
rs377177368 Chr.X: 106632650-106632650 on Build GRCh38
rs747314551 Chr.X: 106632906-106632906 on Build GRCh38
rs113822675 Chr.X: 106632869-106632870 on Build GRCh38
rs570540571 Chr.X: 106633009-106633009 on Build GRCh38
rs763178702 Chr.X: 106632674-106632674 on Build GRCh38
rs769809960 Chr.X: 106632972-106632972 on Build GRCh38
rs766946952 Chr.X: 106632733-106632734 on Build GRCh38
rs774652094 Chr.X: 106633024-106633024 on Build GRCh38
rs983250178 Chr.X: 106632836-106632836 on Build GRCh38
rs950512318 Chr.X: 106632832-106632832 on Build GRCh38
rs775537317 Chr.X: 106632977-106632977 on Build GRCh38
rs777210803 Chr.X: 106632925-106632925 on Build GRCh38
rs778578549 Chr.X: 106632884-106632884 on Build GRCh38
rs201783164 Chr.X: 106633048-106633048 on Build GRCh38
rs771350177 Chr.X: 106632925-106632925 on Build GRCh38
rs770522347 Chr.X: 106632827-106632827 on Build GRCh38
rs1049736992 Chr.X: 106633136-106633136 on Build GRCh38
rs1029314529 Chr.X: 106632845-106632845 on Build GRCh38
rs765331531 Chr.X: 106633054-106633054 on Build GRCh38
rs770285888 Chr.X: 106633124-106633124 on Build GRCh38
rs4039713 Chr.X: 106632864-106632865 on Build GRCh38
rs777244113 Chr.X: 106633091-106633091 on Build GRCh38
rs746611349 Chr.X: 106632721-106632721 on Build GRCh38
rs1045810221 Chr.X: 106633080-106633080 on Build GRCh38
rs758119934 Chr.X: 106632905-106632905 on Build GRCh38
rs776966238 Chr.X: 106632943-106632943 on Build GRCh38
rs749955891 Chr.X: 106633104-106633104 on Build GRCh38
rs752223373 Chr.X: 106633085-106633085 on Build GRCh38
rs35637076 Chr.X: 106632866-106632867 on Build GRCh38
rs754951171 Chr.X: 106633049-106633049 on Build GRCh38
rs768026639 Chr.X: 106632643-106632643 on Build GRCh38
rs200537114 Chr.X: 106633107-106633107 on Build GRCh38
rs753842180 Chr.X: 106632748-106632748 on Build GRCh38

View Full Product Details