Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106918277-106918779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN2
Entrez Gene ID
9075
Gene Name
claudin 2
Gene Aliases
-
UniGene
Hs.522746
Gene Chromosome Location
Chr.X: 106900164-106930861 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171095.1

SNP Detail

SNP ID Chromosome Location
rs1013123551 Chr.X: 106918544-106918544 on Build GRCh38
rs191327348 Chr.X: 106918381-106918381 on Build GRCh38
rs781579044 Chr.X: 106918400-106918400 on Build GRCh38
rs767927136 Chr.X: 106918321-106918321 on Build GRCh38
rs781154153 Chr.X: 106918655-106918655 on Build GRCh38
rs759210574 Chr.X: 106918430-106918430 on Build GRCh38
rs927620472 Chr.X: 106918310-106918310 on Build GRCh38
rs969130770 Chr.X: 106918633-106918633 on Build GRCh38
rs975309865 Chr.X: 106918699-106918699 on Build GRCh38
rs150988328 Chr.X: 106918482-106918482 on Build GRCh38
rs951235861 Chr.X: 106918318-106918318 on Build GRCh38
rs758224177 Chr.X: 106918348-106918349 on Build GRCh38
rs757996708 Chr.X: 106918477-106918477 on Build GRCh38
rs752314939 Chr.X: 106918457-106918457 on Build GRCh38
rs984882495 Chr.X: 106918497-106918497 on Build GRCh38
rs771371843 Chr.X: 106918483-106918483 on Build GRCh38
rs941233929 Chr.X: 106918414-106918414 on Build GRCh38
rs868265725 Chr.X: 106918433-106918433 on Build GRCh38
rs962860627 Chr.X: 106918694-106918694 on Build GRCh38
rs902112410 Chr.X: 106918625-106918625 on Build GRCh38
rs755938708 Chr.X: 106918701-106918701 on Build GRCh38
rs140408996 Chr.X: 106918434-106918434 on Build GRCh38
rs140820041 Chr.X: 106918557-106918557 on Build GRCh38
rs1022051327 Chr.X: 106918749-106918749 on Build GRCh38
rs182735876 Chr.X: 106918471-106918471 on Build GRCh38
rs889494228 Chr.X: 106918716-106918716 on Build GRCh38
rs933980437 Chr.X: 106918440-106918440 on Build GRCh38
rs967941023 Chr.X: 106918425-106918425 on Build GRCh38
rs951039153 Chr.X: 106918648-106918648 on Build GRCh38
rs1036983276 Chr.X: 106918623-106918623 on Build GRCh38
rs923881074 Chr.X: 106918458-106918458 on Build GRCh38
rs1040176449 Chr.X: 106918431-106918431 on Build GRCh38
rs750289241 Chr.X: 106918670-106918670 on Build GRCh38
rs1003705169 Chr.X: 106918637-106918637 on Build GRCh38
rs748173436 Chr.X: 106918416-106918416 on Build GRCh38
rs909363841 Chr.X: 106918526-106918526 on Build GRCh38
rs909826229 Chr.X: 106918690-106918690 on Build GRCh38
rs187024670 Chr.X: 106918659-106918659 on Build GRCh38
rs377475061 Chr.X: 106918588-106918588 on Build GRCh38
rs143589451 Chr.X: 106918479-106918479 on Build GRCh38
rs1006636915 Chr.X: 106918717-106918717 on Build GRCh38
rs989097069 Chr.X: 106918668-106918668 on Build GRCh38

View Full Product Details