Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106929955-106930448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN2
Entrez Gene ID
9075
Gene Name
claudin 2
Gene Aliases
-
UniGene
Hs.522746
Gene Chromosome Location
Chr.X: 106900164-106930861 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171095.1 NM_020384.3

SNP Detail

SNP ID Chromosome Location
rs1035386486 Chr.X: 106930279-106930279 on Build GRCh38
rs775069708 Chr.X: 106930163-106930163 on Build GRCh38
rs926579900 Chr.X: 106930228-106930228 on Build GRCh38
rs749797089 Chr.X: 106930065-106930065 on Build GRCh38
rs184610738 Chr.X: 106930086-106930086 on Build GRCh38
rs1004419212 Chr.X: 106930372-106930372 on Build GRCh38
rs779196711 Chr.X: 106930098-106930098 on Build GRCh38
rs993619322 Chr.X: 106930334-106930334 on Build GRCh38
rs145166443 Chr.X: 106930229-106930229 on Build GRCh38
rs769158456 Chr.X: 106930371-106930371 on Build GRCh38
rs1044721618 Chr.X: 106930257-106930257 on Build GRCh38
rs917799064 Chr.X: 106930167-106930167 on Build GRCh38
rs1034844997 Chr.X: 106929993-106929998 on Build GRCh38
rs745435515 Chr.X: 106930283-106930283 on Build GRCh38
rs987916679 Chr.X: 106930166-106930166 on Build GRCh38
rs373807733 Chr.X: 106930381-106930381 on Build GRCh38
rs1052185255 Chr.X: 106930203-106930203 on Build GRCh38
rs897892712 Chr.X: 106930275-106930275 on Build GRCh38
rs878973134 Chr.X: 106930236-106930236 on Build GRCh38
rs982037199 Chr.X: 106929977-106929977 on Build GRCh38
rs1018323664 Chr.X: 106930081-106930081 on Build GRCh38
rs921283334 Chr.X: 106930122-106930122 on Build GRCh38
rs904886092 Chr.X: 106930260-106930260 on Build GRCh38
rs934216616 Chr.X: 106930146-106930146 on Build GRCh38
rs141297568 Chr.X: 106930136-106930136 on Build GRCh38
rs771337109 Chr.X: 106930347-106930347 on Build GRCh38
rs186720703 Chr.X: 106930391-106930391 on Build GRCh38
rs1010676925 Chr.X: 106930216-106930216 on Build GRCh38
rs949321986 Chr.X: 106930193-106930193 on Build GRCh38
rs956485078 Chr.X: 106930150-106930150 on Build GRCh38
rs951313095 Chr.X: 106930032-106930032 on Build GRCh38
rs1030521600 Chr.X: 106930027-106930027 on Build GRCh38
rs982884104 Chr.X: 106930046-106930046 on Build GRCh38
rs777233825 Chr.X: 106930076-106930077 on Build GRCh38
rs993651568 Chr.X: 106929989-106929989 on Build GRCh38
rs924786790 Chr.X: 106930152-106930152 on Build GRCh38
rs941456311 Chr.X: 106930066-106930066 on Build GRCh38
rs1007842765 Chr.X: 106930076-106930076 on Build GRCh38
rs1037732398 Chr.X: 106930269-106930269 on Build GRCh38
rs1044998511 Chr.X: 106930220-106930220 on Build GRCh38
rs951237979 Chr.X: 106930329-106930329 on Build GRCh38
rs780022444 Chr.X: 106930423-106930423 on Build GRCh38
rs893519288 Chr.X: 106930208-106930208 on Build GRCh38
rs1051814528 Chr.X: 106930151-106930151 on Build GRCh38
rs1016771138 Chr.X: 106930392-106930392 on Build GRCh38
rs778318354 Chr.X: 106929997-106929997 on Build GRCh38
rs147829672 Chr.X: 106930111-106930111 on Build GRCh38
rs955125195 Chr.X: 106930046-106930046 on Build GRCh38

View Full Product Details