Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181720123-181720634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
UniGene
Hs.437444
Gene Chromosome Location
Chr.1: 181317712-181808084 on Build GRCh38
Transcripts
NM_000721.3 NM_001205293.1 NM_001205294.1 XM_017002243.1 XM_017002244.1 XM_017002245.1 XM_017002246.1 XM_017002247.1 XM_017002248.1 XM_017002249.1 XM_017002250.1 XM_017002251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4900020 c.1800G>A p.M600I Chr.1: 181720254-181720254 on Build GRCh38
5208549 c.1636T>C p.S546P Chr.1: 181720237-181720237 on Build GRCh38
3863777 c.1828C>T p.L610F Chr.1: 181720282-181720282 on Build GRCh38
1730881 c.1862G>T p.G621V Chr.1: 181720316-181720316 on Build GRCh38
6575322 c.1689C>A p.L563L Chr.1: 181720290-181720290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034824468 Chr.1: 181720452-181720452 on Build GRCh38
rs986236908 Chr.1: 181720178-181720178 on Build GRCh38
rs754092700 Chr.1: 181720474-181720474 on Build GRCh38
rs763378345 Chr.1: 181720159-181720159 on Build GRCh38
rs773898855 Chr.1: 181720169-181720177 on Build GRCh38
rs932173165 Chr.1: 181720152-181720152 on Build GRCh38
rs779331307 Chr.1: 181720389-181720389 on Build GRCh38
rs560597086 Chr.1: 181720366-181720366 on Build GRCh38
rs921007944 Chr.1: 181720151-181720151 on Build GRCh38
rs907262983 Chr.1: 181720494-181720494 on Build GRCh38
rs369480285 Chr.1: 181720341-181720341 on Build GRCh38
rs866685114 Chr.1: 181720254-181720254 on Build GRCh38
rs768348854 Chr.1: 181720520-181720520 on Build GRCh38
rs769996689 Chr.1: 181720352-181720352 on Build GRCh38
rs745432781 Chr.1: 181720447-181720447 on Build GRCh38
rs865996425 Chr.1: 181720312-181720312 on Build GRCh38
rs542490130 Chr.1: 181720149-181720149 on Build GRCh38
rs959290722 Chr.1: 181720161-181720161 on Build GRCh38
rs962719638 Chr.1: 181720495-181720495 on Build GRCh38
rs552457845 Chr.1: 181720410-181720410 on Build GRCh38
rs760975849 Chr.1: 181720222-181720222 on Build GRCh38
rs532066763 Chr.1: 181720430-181720430 on Build GRCh38
rs1043709803 Chr.1: 181720586-181720586 on Build GRCh38
rs577349639 Chr.1: 181720478-181720478 on Build GRCh38
rs1033500213 Chr.1: 181720527-181720527 on Build GRCh38
rs752102506 Chr.1: 181720191-181720191 on Build GRCh38
rs568489899 Chr.1: 181720538-181720538 on Build GRCh38
rs548183674 Chr.1: 181720578-181720578 on Build GRCh38
rs373361032 Chr.1: 181720294-181720294 on Build GRCh38
rs762337372 Chr.1: 181720209-181720209 on Build GRCh38
rs768854440 Chr.1: 181720381-181720381 on Build GRCh38
rs959492355 Chr.1: 181720537-181720537 on Build GRCh38
rs879655024 Chr.1: 181720188-181720188 on Build GRCh38
rs746104157 Chr.1: 181720348-181720348 on Build GRCh38
rs936252158 Chr.1: 181720417-181720417 on Build GRCh38
rs1055155907 Chr.1: 181720438-181720438 on Build GRCh38
rs889962931 Chr.1: 181720412-181720412 on Build GRCh38
rs961325603 Chr.1: 181720577-181720577 on Build GRCh38
rs764608071 Chr.1: 181720170-181720170 on Build GRCh38
rs775594936 Chr.1: 181720156-181720156 on Build GRCh38
rs769263659 Chr.1: 181720449-181720449 on Build GRCh38
rs145289624 Chr.1: 181720421-181720421 on Build GRCh38
rs1028507267 Chr.1: 181720510-181720510 on Build GRCh38
rs1017297736 Chr.1: 181720550-181720550 on Build GRCh38
rs767963797 Chr.1: 181720215-181720215 on Build GRCh38
rs550292890 Chr.1: 181720490-181720490 on Build GRCh38
rs946731794 Chr.1: 181720582-181720582 on Build GRCh38
rs953765323 Chr.1: 181720528-181720528 on Build GRCh38
rs763153720 Chr.1: 181720373-181720373 on Build GRCh38
rs374286518 Chr.1: 181720221-181720221 on Build GRCh38
rs764352459 Chr.1: 181720296-181720296 on Build GRCh38
rs1033531217 Chr.1: 181720148-181720148 on Build GRCh38
rs933742258 Chr.1: 181720382-181720382 on Build GRCh38
rs749175126 Chr.1: 181720496-181720496 on Build GRCh38
rs946731265 Chr.1: 181720370-181720370 on Build GRCh38
rs191216596 Chr.1: 181720576-181720576 on Build GRCh38
rs374784506 Chr.1: 181720304-181720304 on Build GRCh38
rs778291283 Chr.1: 181720261-181720261 on Build GRCh38
rs746915512 Chr.1: 181720297-181720297 on Build GRCh38
rs116114878 Chr.1: 181720384-181720384 on Build GRCh38
rs752193507 Chr.1: 181720267-181720267 on Build GRCh38
rs770758448 Chr.1: 181720335-181720335 on Build GRCh38
rs939720420 Chr.1: 181720326-181720326 on Build GRCh38
rs768016860 Chr.1: 181720385-181720385 on Build GRCh38

View Full Product Details