Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182381567-182382081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUL
Entrez Gene ID
2752
Gene Name
glutamate-ammonia ligase
Gene Aliases
GLNS, GS, PIG43, PIG59
UniGene
Hs.132016
Gene Chromosome Location
Chr.1: 182378093-182392206 on Build GRCh38
Transcripts
NM_001033044.3 NM_001033056.3 NM_002065.6

SNP Detail

SNP ID Chromosome Location
rs960123809 Chr.1: 182381991-182381991 on Build GRCh38
rs968629850 Chr.1: 182382027-182382027 on Build GRCh38
rs1055158749 Chr.1: 182381801-182381801 on Build GRCh38
rs2296523 Chr.1: 182381761-182381761 on Build GRCh38
rs895290811 Chr.1: 182381817-182381817 on Build GRCh38
rs535513436 Chr.1: 182381813-182381813 on Build GRCh38
rs941536233 Chr.1: 182381819-182381819 on Build GRCh38
rs368774739 Chr.1: 182381668-182381668 on Build GRCh38
rs1015966219 Chr.1: 182381791-182381791 on Build GRCh38
rs766506428 Chr.1: 182381977-182381978 on Build GRCh38
rs903306108 Chr.1: 182381670-182381670 on Build GRCh38
rs1049949036 Chr.1: 182381918-182381918 on Build GRCh38
rs916657978 Chr.1: 182381953-182381953 on Build GRCh38
rs768648311 Chr.1: 182381927-182381927 on Build GRCh38
rs752788371 Chr.1: 182381631-182381631 on Build GRCh38
rs981352674 Chr.1: 182381928-182381928 on Build GRCh38
rs939425181 Chr.1: 182381798-182381798 on Build GRCh38
rs945356932 Chr.1: 182381768-182381768 on Build GRCh38
rs576317186 Chr.1: 182382004-182382004 on Build GRCh38
rs890819500 Chr.1: 182381850-182381850 on Build GRCh38
rs140748780 Chr.1: 182382040-182382040 on Build GRCh38
rs985238276 Chr.1: 182381680-182381680 on Build GRCh38
rs142906434 Chr.1: 182381778-182381778 on Build GRCh38
rs971891161 Chr.1: 182381796-182381796 on Build GRCh38
rs1043112165 Chr.1: 182381835-182381836 on Build GRCh38
rs188390414 Chr.1: 182382048-182382048 on Build GRCh38
rs939371156 Chr.1: 182381946-182381946 on Build GRCh38
rs752049541 Chr.1: 182381763-182381763 on Build GRCh38
rs2296522 Chr.1: 182381774-182381774 on Build GRCh38
rs1038582398 Chr.1: 182381822-182381822 on Build GRCh38
rs991800541 Chr.1: 182381802-182381802 on Build GRCh38
rs886045599 Chr.1: 182381801-182381801 on Build GRCh38
rs537395125 Chr.1: 182382005-182382005 on Build GRCh38
rs927948541 Chr.1: 182381942-182381942 on Build GRCh38
rs774275913 Chr.1: 182382031-182382031 on Build GRCh38
rs765260837 Chr.1: 182381599-182381599 on Build GRCh38
rs77070600 Chr.1: 182381911-182381911 on Build GRCh38
rs555319487 Chr.1: 182382006-182382006 on Build GRCh38
rs745540918 Chr.1: 182381806-182381806 on Build GRCh38
rs748818045 Chr.1: 182381887-182381887 on Build GRCh38
rs886045600 Chr.1: 182381942-182381942 on Build GRCh38
rs990833306 Chr.1: 182381948-182381948 on Build GRCh38
rs183655903 Chr.1: 182381841-182381841 on Build GRCh38
rs191511731 Chr.1: 182381834-182381834 on Build GRCh38
rs941496181 Chr.1: 182381979-182381979 on Build GRCh38
rs957451439 Chr.1: 182381721-182381721 on Build GRCh38
rs187700618 Chr.1: 182381657-182381657 on Build GRCh38
rs369097343 Chr.1: 182382037-182382038 on Build GRCh38
rs1018455029 Chr.1: 182381888-182381888 on Build GRCh38
rs1020210290 Chr.1: 182381743-182381743 on Build GRCh38
rs961805228 Chr.1: 182381793-182381793 on Build GRCh38
rs928463393 Chr.1: 182381799-182381800 on Build GRCh38
rs552417488 Chr.1: 182381685-182381685 on Build GRCh38
rs1018102070 Chr.1: 182381694-182381694 on Build GRCh38
rs146613976 Chr.1: 182381808-182381808 on Build GRCh38
rs779481807 Chr.1: 182381837-182381837 on Build GRCh38
rs759018151 Chr.1: 182381700-182381700 on Build GRCh38
rs140277335 Chr.1: 182381889-182381889 on Build GRCh38
rs577652875 Chr.1: 182381773-182381773 on Build GRCh38
rs934743402 Chr.1: 182381883-182381883 on Build GRCh38
rs1055773294 Chr.1: 182381619-182381619 on Build GRCh38
rs531293807 Chr.1: 182381769-182381769 on Build GRCh38
rs913149476 Chr.1: 182381762-182381762 on Build GRCh38
rs146420457 Chr.1: 182382007-182382007 on Build GRCh38
rs993664066 Chr.1: 182381965-182381965 on Build GRCh38
rs993837516 Chr.1: 182381611-182381611 on Build GRCh38
rs990391065 Chr.1: 182381907-182381907 on Build GRCh38

View Full Product Details