Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183648500-183649011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC4
Entrez Gene ID
403314
Gene Name
apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4
Gene Aliases
C1orf169
UniGene
Hs.97335
Gene Chromosome Location
Chr.1: 183646275-183653313 on Build GRCh38
Transcripts
NM_203454.2 NM_001297669.1 NM_001297670.1 NM_015149.4 XM_011509339.2 XM_011509341.1 XM_011509342.2

Gene Symbol
RGL1
Entrez Gene ID
23179
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Gene Aliases
RGL
UniGene
Hs.497148
Gene Chromosome Location
Chr.1: 183636047-183928551 on Build GRCh38
Transcripts
NM_203454.2 NM_001297669.1 NM_001297670.1 NM_015149.4 XM_011509339.2 XM_011509341.1 XM_011509342.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060433 c.41C>T p.T14I Chr.1: 183648741-183648741 on Build GRCh38
6123487 c.60C>A p.Y20* Chr.1: 183648722-183648722 on Build GRCh38
1337044 c.263C>T p.T88I Chr.1: 183648519-183648519 on Build GRCh38
4437661 c.78C>T p.L26L Chr.1: 183648704-183648704 on Build GRCh38
900837 c.203T>G p.F68C Chr.1: 183648579-183648579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977275340 Chr.1: 183648888-183648888 on Build GRCh38
rs1053536503 Chr.1: 183648661-183648661 on Build GRCh38
rs78130638 Chr.1: 183648981-183648981 on Build GRCh38
rs770802488 Chr.1: 183648729-183648729 on Build GRCh38
rs950811725 Chr.1: 183648910-183648910 on Build GRCh38
rs963956451 Chr.1: 183648801-183648801 on Build GRCh38
rs151221520 Chr.1: 183648680-183648680 on Build GRCh38
rs747300943 Chr.1: 183648572-183648572 on Build GRCh38
rs763431906 Chr.1: 183648645-183648645 on Build GRCh38
rs1030015344 Chr.1: 183648743-183648743 on Build GRCh38
rs762009746 Chr.1: 183648671-183648671 on Build GRCh38
rs776474653 Chr.1: 183648660-183648660 on Build GRCh38
rs776661029 Chr.1: 183648588-183648588 on Build GRCh38
rs761340418 Chr.1: 183648647-183648648 on Build GRCh38
rs569838577 Chr.1: 183648913-183648913 on Build GRCh38
rs746049276 Chr.1: 183648769-183648769 on Build GRCh38
rs370657788 Chr.1: 183648571-183648571 on Build GRCh38
rs781510132 Chr.1: 183648738-183648738 on Build GRCh38
rs192632617 Chr.1: 183648847-183648847 on Build GRCh38
rs201259808 Chr.1: 183648662-183648662 on Build GRCh38
rs183310381 Chr.1: 183648985-183648985 on Build GRCh38
rs1026359882 Chr.1: 183648873-183648873 on Build GRCh38
rs16861394 Chr.1: 183648558-183648558 on Build GRCh38
rs769161091 Chr.1: 183648788-183648788 on Build GRCh38
rs753574253 Chr.1: 183648797-183648797 on Build GRCh38
rs1023762764 Chr.1: 183648684-183648684 on Build GRCh38
rs562884646 Chr.1: 183648656-183648656 on Build GRCh38
rs750583172 Chr.1: 183648826-183648826 on Build GRCh38
rs951485706 Chr.1: 183648792-183648792 on Build GRCh38
rs202076412 Chr.1: 183648587-183648587 on Build GRCh38
rs529403451 Chr.1: 183648531-183648531 on Build GRCh38
rs1056765312 Chr.1: 183648974-183648974 on Build GRCh38
rs759095376 Chr.1: 183648799-183648799 on Build GRCh38
rs376258698 Chr.1: 183648553-183648553 on Build GRCh38
rs992762314 Chr.1: 183648912-183648912 on Build GRCh38
rs777786538 Chr.1: 183648719-183648719 on Build GRCh38
rs555343776 Chr.1: 183648980-183648980 on Build GRCh38
rs764868306 Chr.1: 183648800-183648800 on Build GRCh38
rs12088609 Chr.1: 183648930-183648930 on Build GRCh38
rs144155004 Chr.1: 183648704-183648704 on Build GRCh38
rs544160727 Chr.1: 183648746-183648746 on Build GRCh38
rs766038265 Chr.1: 183648667-183648667 on Build GRCh38
rs757924008 Chr.1: 183648825-183648825 on Build GRCh38
rs980702019 Chr.1: 183648946-183648946 on Build GRCh38
rs763522170 Chr.1: 183648778-183648778 on Build GRCh38
rs778262711 Chr.1: 183648676-183648676 on Build GRCh38
rs947191629 Chr.1: 183648861-183648861 on Build GRCh38
rs551756459 Chr.1: 183648846-183648846 on Build GRCh38
rs765193470 Chr.1: 183648613-183648613 on Build GRCh38
rs898778620 Chr.1: 183648989-183648989 on Build GRCh38
rs767739378 Chr.1: 183648795-183648795 on Build GRCh38
rs776107825 Chr.1: 183648641-183648641 on Build GRCh38
rs774786631 Chr.1: 183648794-183648794 on Build GRCh38
rs746814359 Chr.1: 183648728-183648728 on Build GRCh38
rs752259873 Chr.1: 183648811-183648811 on Build GRCh38
rs141035551 Chr.1: 183648770-183648770 on Build GRCh38
rs765468557 Chr.1: 183648807-183648808 on Build GRCh38
rs775423406 Chr.1: 183648771-183648771 on Build GRCh38
rs753388975 Chr.1: 183648675-183648675 on Build GRCh38
rs948449902 Chr.1: 183648984-183648984 on Build GRCh38
rs759685764 Chr.1: 183648600-183648600 on Build GRCh38
rs368111873 Chr.1: 183648649-183648649 on Build GRCh38
rs182486585 Chr.1: 183648660-183648660 on Build GRCh38

View Full Product Details