Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183936129-183936640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
UniGene
Hs.387995
Gene Chromosome Location
Chr.1: 183929662-184051612 on Build GRCh38
Transcripts
NM_001303420.1 NM_001303421.1 NM_015101.3 XM_011509332.2

SNP Detail

SNP ID Chromosome Location
rs574584623 Chr.1: 183936224-183936224 on Build GRCh38
rs759016076 Chr.1: 183936206-183936219 on Build GRCh38
rs376004549 Chr.1: 183936163-183936163 on Build GRCh38
rs539775143 Chr.1: 183936532-183936532 on Build GRCh38
rs1040594174 Chr.1: 183936553-183936553 on Build GRCh38
rs760334750 Chr.1: 183936202-183936202 on Build GRCh38
rs1030136280 Chr.1: 183936278-183936278 on Build GRCh38
rs200540977 Chr.1: 183936425-183936432 on Build GRCh38
rs559172535 Chr.1: 183936154-183936154 on Build GRCh38
rs1055727088 Chr.1: 183936192-183936192 on Build GRCh38
rs183697010 Chr.1: 183936218-183936218 on Build GRCh38
rs897736693 Chr.1: 183936249-183936249 on Build GRCh38
rs71657827 Chr.1: 183936351-183936352 on Build GRCh38
rs151252912 Chr.1: 183936488-183936489 on Build GRCh38
rs12023991 Chr.1: 183936343-183936343 on Build GRCh38
rs997382756 Chr.1: 183936275-183936275 on Build GRCh38
rs542269529 Chr.1: 183936229-183936229 on Build GRCh38
rs893490069 Chr.1: 183936234-183936234 on Build GRCh38
rs1055834270 Chr.1: 183936596-183936596 on Build GRCh38
rs149271313 Chr.1: 183936152-183936152 on Build GRCh38
rs753093438 Chr.1: 183936507-183936507 on Build GRCh38
rs946355735 Chr.1: 183936260-183936260 on Build GRCh38
rs942063931 Chr.1: 183936605-183936605 on Build GRCh38
rs1042033905 Chr.1: 183936514-183936515 on Build GRCh38
rs924230076 Chr.1: 183936525-183936525 on Build GRCh38
rs1019525647 Chr.1: 183936291-183936291 on Build GRCh38
rs575661785 Chr.1: 183936473-183936473 on Build GRCh38
rs953110080 Chr.1: 183936309-183936309 on Build GRCh38
rs1027718951 Chr.1: 183936320-183936320 on Build GRCh38
rs1015278180 Chr.1: 183936248-183936248 on Build GRCh38
rs560289185 Chr.1: 183936300-183936300 on Build GRCh38
rs907333853 Chr.1: 183936188-183936188 on Build GRCh38
rs534225403 Chr.1: 183936153-183936153 on Build GRCh38
rs3841431 Chr.1: 183936355-183936356 on Build GRCh38
rs996668793 Chr.1: 183936312-183936312 on Build GRCh38
rs369763217 Chr.1: 183936429-183936429 on Build GRCh38
rs1045873525 Chr.1: 183936187-183936187 on Build GRCh38
rs913514226 Chr.1: 183936399-183936399 on Build GRCh38
rs377335447 Chr.1: 183936383-183936383 on Build GRCh38
rs961580134 Chr.1: 183936359-183936359 on Build GRCh38
rs900637340 Chr.1: 183936305-183936305 on Build GRCh38
rs928769593 Chr.1: 183936502-183936502 on Build GRCh38
rs887847941 Chr.1: 183936600-183936600 on Build GRCh38
rs902755985 Chr.1: 183936276-183936276 on Build GRCh38
rs565170629 Chr.1: 183936398-183936398 on Build GRCh38
rs765950038 Chr.1: 183936324-183936324 on Build GRCh38
rs990988371 Chr.1: 183936418-183936418 on Build GRCh38
rs1042649227 Chr.1: 183936266-183936266 on Build GRCh38
rs952052700 Chr.1: 183936327-183936327 on Build GRCh38
rs572377599 Chr.1: 183936339-183936339 on Build GRCh38
rs946231566 Chr.1: 183936183-183936183 on Build GRCh38
rs577060373 Chr.1: 183936189-183936189 on Build GRCh38
rs538029396 Chr.1: 183936196-183936196 on Build GRCh38
rs374486344 Chr.1: 183936415-183936415 on Build GRCh38
rs186964186 Chr.1: 183936612-183936612 on Build GRCh38
rs946315037 Chr.1: 183936401-183936401 on Build GRCh38
rs773769004 Chr.1: 183936309-183936309 on Build GRCh38
rs937834933 Chr.1: 183936413-183936413 on Build GRCh38
rs868396777 Chr.1: 183936231-183936231 on Build GRCh38
rs984058493 Chr.1: 183936151-183936151 on Build GRCh38

View Full Product Details