Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183937996-183938510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
UniGene
Hs.387995
Gene Chromosome Location
Chr.1: 183929662-184051612 on Build GRCh38
Transcripts
NM_001303420.1 NM_001303421.1 NM_015101.3 XM_011509332.2 XM_011509334.2 XM_011509335.2 XM_011509336.2 XM_011509337.2 XM_011509338.2 XM_017000750.1 XM_017000751.1

SNP Detail

SNP ID Chromosome Location
rs968832419 Chr.1: 183938369-183938369 on Build GRCh38
rs116234834 Chr.1: 183938440-183938440 on Build GRCh38
rs989502469 Chr.1: 183938033-183938033 on Build GRCh38
rs990622183 Chr.1: 183938350-183938350 on Build GRCh38
rs900586378 Chr.1: 183938296-183938296 on Build GRCh38
rs977154968 Chr.1: 183938387-183938387 on Build GRCh38
rs747911674 Chr.1: 183938298-183938298 on Build GRCh38
rs374253430 Chr.1: 183938339-183938339 on Build GRCh38
rs749519738 Chr.1: 183938136-183938136 on Build GRCh38
rs933241558 Chr.1: 183938417-183938417 on Build GRCh38
rs189874182 Chr.1: 183938321-183938321 on Build GRCh38
rs36161033 Chr.1: 183938212-183938212 on Build GRCh38
rs923247855 Chr.1: 183938397-183938397 on Build GRCh38
rs10664802 Chr.1: 183938282-183938283 on Build GRCh38
rs551876258 Chr.1: 183938173-183938173 on Build GRCh38
rs990610560 Chr.1: 183938103-183938103 on Build GRCh38
rs924632053 Chr.1: 183938100-183938100 on Build GRCh38
rs3828027 Chr.1: 183938118-183938118 on Build GRCh38
rs3841432 Chr.1: 183938198-183938198 on Build GRCh38
rs927871860 Chr.1: 183938068-183938068 on Build GRCh38
rs75067855 Chr.1: 183938211-183938211 on Build GRCh38
rs151194611 Chr.1: 183938081-183938081 on Build GRCh38
rs57324076 Chr.1: 183938210-183938210 on Build GRCh38
rs369173689 Chr.1: 183938124-183938125 on Build GRCh38
rs916367806 Chr.1: 183938106-183938106 on Build GRCh38
rs1037432379 Chr.1: 183938205-183938206 on Build GRCh38
rs1035970500 Chr.1: 183938259-183938259 on Build GRCh38
rs79919750 Chr.1: 183938212-183938212 on Build GRCh38
rs60413881 Chr.1: 183938374-183938374 on Build GRCh38
rs977949384 Chr.1: 183938063-183938063 on Build GRCh38
rs1009110794 Chr.1: 183938274-183938274 on Build GRCh38
rs765619859 Chr.1: 183938212-183938212 on Build GRCh38
rs141136994 Chr.1: 183938281-183938282 on Build GRCh38
rs968779891 Chr.1: 183938062-183938062 on Build GRCh38
rs772253490 Chr.1: 183938335-183938335 on Build GRCh38
rs141272146 Chr.1: 183938210-183938212 on Build GRCh38
rs185071329 Chr.1: 183938310-183938310 on Build GRCh38
rs962510112 Chr.1: 183938398-183938398 on Build GRCh38
rs1050747764 Chr.1: 183938485-183938485 on Build GRCh38
rs201589079 Chr.1: 183938209-183938209 on Build GRCh38
rs535233856 Chr.1: 183938128-183938128 on Build GRCh38
rs796255256 Chr.1: 183938210-183938212 on Build GRCh38
rs888918045 Chr.1: 183938180-183938180 on Build GRCh38
rs386368953 Chr.1: 183938283-183938284 on Build GRCh38
rs1034939176 Chr.1: 183938342-183938342 on Build GRCh38
rs771935919 Chr.1: 183938334-183938334 on Build GRCh38
rs879063783 Chr.1: 183938210-183938210 on Build GRCh38
rs929654419 Chr.1: 183938464-183938464 on Build GRCh38
rs776514545 Chr.1: 183938198-183938199 on Build GRCh38
rs960603510 Chr.1: 183938346-183938346 on Build GRCh38
rs199853407 Chr.1: 183938227-183938227 on Build GRCh38
rs376972155 Chr.1: 183938338-183938338 on Build GRCh38
rs879335927 Chr.1: 183938113-183938113 on Build GRCh38
rs993559075 Chr.1: 183938209-183938210 on Build GRCh38
rs890694859 Chr.1: 183938178-183938178 on Build GRCh38
rs181294624 Chr.1: 183938147-183938147 on Build GRCh38
rs1039576988 Chr.1: 183938286-183938286 on Build GRCh38
rs869106241 Chr.1: 183938287-183938288 on Build GRCh38
rs3814332 Chr.1: 183938157-183938157 on Build GRCh38
rs570078143 Chr.1: 183938196-183938196 on Build GRCh38
rs978827326 Chr.1: 183938371-183938371 on Build GRCh38
rs939210774 Chr.1: 183938085-183938085 on Build GRCh38
rs188479720 Chr.1: 183938055-183938055 on Build GRCh38
rs397982224 Chr.1: 183938286-183938287 on Build GRCh38
rs200622927 Chr.1: 183938309-183938309 on Build GRCh38
rs956087888 Chr.1: 183938376-183938376 on Build GRCh38
rs1022306570 Chr.1: 183938327-183938327 on Build GRCh38
rs974201256 Chr.1: 183938423-183938423 on Build GRCh38
rs1023090505 Chr.1: 183938357-183938357 on Build GRCh38
rs1022675895 Chr.1: 183938040-183938040 on Build GRCh38

View Full Product Details