Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 184621331-184621837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf21
Entrez Gene ID
81563
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
PIG13
UniGene
Hs.497159
Gene Chromosome Location
Chr.1: 184387016-184629021 on Build GRCh38
Transcripts
NM_030806.3

SNP Detail

SNP ID Chromosome Location
rs1043606896 Chr.1: 184621669-184621669 on Build GRCh38
rs752680487 Chr.1: 184621535-184621535 on Build GRCh38
rs576428554 Chr.1: 184621752-184621752 on Build GRCh38
rs998456463 Chr.1: 184621481-184621481 on Build GRCh38
rs547514000 Chr.1: 184621572-184621572 on Build GRCh38
rs147273699 Chr.1: 184621798-184621798 on Build GRCh38
rs890082142 Chr.1: 184621808-184621808 on Build GRCh38
rs943632519 Chr.1: 184621619-184621619 on Build GRCh38
rs143599854 Chr.1: 184621624-184621624 on Build GRCh38
rs139681744 Chr.1: 184621533-184621533 on Build GRCh38
rs537309537 Chr.1: 184621790-184621790 on Build GRCh38
rs1022806445 Chr.1: 184621515-184621515 on Build GRCh38
rs1007533179 Chr.1: 184621816-184621816 on Build GRCh38
rs998751211 Chr.1: 184621413-184621413 on Build GRCh38
rs981584342 Chr.1: 184621534-184621534 on Build GRCh38
rs575628167 Chr.1: 184621555-184621555 on Build GRCh38
rs934282956 Chr.1: 184621704-184621704 on Build GRCh38
rs891771974 Chr.1: 184621454-184621454 on Build GRCh38
rs565627300 Chr.1: 184621580-184621580 on Build GRCh38
rs1051201720 Chr.1: 184621374-184621374 on Build GRCh38
rs902947081 Chr.1: 184621685-184621685 on Build GRCh38
rs925094668 Chr.1: 184621730-184621730 on Build GRCh38
rs1012877757 Chr.1: 184621791-184621791 on Build GRCh38
rs1007090919 Chr.1: 184621520-184621520 on Build GRCh38
rs1012719982 Chr.1: 184621477-184621477 on Build GRCh38
rs35382132 Chr.1: 184621763-184621764 on Build GRCh38
rs891803401 Chr.1: 184621743-184621743 on Build GRCh38
rs1029880034 Chr.1: 184621500-184621500 on Build GRCh38
rs1050895269 Chr.1: 184621667-184621667 on Build GRCh38
rs893070421 Chr.1: 184621432-184621432 on Build GRCh38
rs1054129202 Chr.1: 184621740-184621740 on Build GRCh38
rs960341830 Chr.1: 184621591-184621591 on Build GRCh38
rs184061451 Chr.1: 184621625-184621625 on Build GRCh38
rs774409964 Chr.1: 184621395-184621395 on Build GRCh38
rs1032902684 Chr.1: 184621415-184621415 on Build GRCh38
rs909441043 Chr.1: 184621601-184621601 on Build GRCh38
rs933730190 Chr.1: 184621654-184621654 on Build GRCh38
rs907063193 Chr.1: 184621815-184621815 on Build GRCh38
rs936111901 Chr.1: 184621731-184621731 on Build GRCh38
rs984774378 Chr.1: 184621567-184621567 on Build GRCh38
rs376076759 Chr.1: 184621712-184621712 on Build GRCh38

View Full Product Details