Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111248812-111249321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN6
Entrez Gene ID
827
Gene Name
calpain 6
Gene Aliases
CANPX, CAPNX, CalpM, DJ914P14.1
UniGene
Hs.496593
Gene Chromosome Location
Chr.X: 111245099-111270546 on Build GRCh38
Transcripts
NM_014289.3

SNP Detail

SNP ID Chromosome Location
rs374187461 Chr.X: 111248925-111248925 on Build GRCh38
rs772216309 Chr.X: 111248927-111248927 on Build GRCh38
rs753775981 Chr.X: 111249242-111249242 on Build GRCh38
rs975305637 Chr.X: 111248856-111248856 on Build GRCh38
rs761817849 Chr.X: 111249194-111249194 on Build GRCh38
rs36011386 Chr.X: 111248937-111248937 on Build GRCh38
rs778090365 Chr.X: 111248922-111248922 on Build GRCh38
rs773560608 Chr.X: 111248958-111248959 on Build GRCh38
rs760960972 Chr.X: 111249100-111249100 on Build GRCh38
rs748323677 Chr.X: 111248922-111248922 on Build GRCh38
rs759479751 Chr.X: 111248954-111248954 on Build GRCh38
rs923913114 Chr.X: 111248985-111248985 on Build GRCh38
rs951641241 Chr.X: 111249203-111249203 on Build GRCh38
rs954389542 Chr.X: 111248881-111248881 on Build GRCh38
rs754512849 Chr.X: 111249024-111249024 on Build GRCh38
rs758015443 Chr.X: 111249052-111249052 on Build GRCh38
rs933809274 Chr.X: 111248916-111248916 on Build GRCh38
rs939314988 Chr.X: 111249022-111249022 on Build GRCh38
rs17880042 Chr.X: 111249111-111249111 on Build GRCh38
rs778360866 Chr.X: 111248894-111248894 on Build GRCh38
rs760708333 Chr.X: 111248984-111248984 on Build GRCh38
rs376298299 Chr.X: 111248960-111248960 on Build GRCh38
rs778471859 Chr.X: 111249025-111249025 on Build GRCh38
rs1004117492 Chr.X: 111249167-111249167 on Build GRCh38
rs779088070 Chr.X: 111248891-111248892 on Build GRCh38
rs747327631 Chr.X: 111249096-111249096 on Build GRCh38
rs771389671 Chr.X: 111248930-111248930 on Build GRCh38
rs143524909 Chr.X: 111248994-111248994 on Build GRCh38
rs1012559995 Chr.X: 111249073-111249073 on Build GRCh38
rs767525041 Chr.X: 111249210-111249210 on Build GRCh38
rs139917382 Chr.X: 111248981-111248981 on Build GRCh38
rs769867424 Chr.X: 111248885-111248885 on Build GRCh38
rs915311560 Chr.X: 111248902-111248902 on Build GRCh38
rs189403027 Chr.X: 111249030-111249030 on Build GRCh38
rs905479043 Chr.X: 111249112-111249112 on Build GRCh38
rs753391368 Chr.X: 111249010-111249010 on Build GRCh38
rs772482333 Chr.X: 111248920-111248920 on Build GRCh38
rs763257826 Chr.X: 111248979-111248979 on Build GRCh38
rs775646239 Chr.X: 111248965-111248965 on Build GRCh38
rs370514418 Chr.X: 111248918-111248918 on Build GRCh38
rs1056819614 Chr.X: 111249090-111249090 on Build GRCh38
rs1012496425 Chr.X: 111249157-111249157 on Build GRCh38
rs113109941 Chr.X: 111249136-111249136 on Build GRCh38
rs766120221 Chr.X: 111249146-111249146 on Build GRCh38
rs778143802 Chr.X: 111249087-111249087 on Build GRCh38
rs887544165 Chr.X: 111249228-111249228 on Build GRCh38
rs750201143 Chr.X: 111248982-111248982 on Build GRCh38

View Full Product Details