Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111295936-111296456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs774218828 Chr.X: 111296198-111296198 on Build GRCh38
rs751638117 Chr.X: 111296246-111296246 on Build GRCh38
rs912653236 Chr.X: 111296348-111296348 on Build GRCh38
rs921261540 Chr.X: 111296402-111296402 on Build GRCh38
rs757285791 Chr.X: 111296248-111296248 on Build GRCh38
rs763233535 Chr.X: 111296197-111296197 on Build GRCh38
rs781270404 Chr.X: 111296258-111296258 on Build GRCh38
rs1024690942 Chr.X: 111295967-111295967 on Build GRCh38
rs944100131 Chr.X: 111296361-111296361 on Build GRCh38
rs930178212 Chr.X: 111296412-111296412 on Build GRCh38
rs951545201 Chr.X: 111296317-111296317 on Build GRCh38
rs759572899 Chr.X: 111296006-111296006 on Build GRCh38
rs750175756 Chr.X: 111296358-111296358 on Build GRCh38
rs975853558 Chr.X: 111296368-111296368 on Build GRCh38
rs752440362 Chr.X: 111296068-111296068 on Build GRCh38
rs1027590507 Chr.X: 111296202-111296202 on Build GRCh38
rs982920055 Chr.X: 111296335-111296335 on Build GRCh38
rs1033351496 Chr.X: 111296118-111296118 on Build GRCh38
rs959250194 Chr.X: 111296144-111296144 on Build GRCh38
rs925748856 Chr.X: 111296079-111296079 on Build GRCh38
rs767370020 Chr.X: 111296305-111296305 on Build GRCh38
rs764283629 Chr.X: 111296245-111296245 on Build GRCh38
rs768605282 Chr.X: 111296028-111296028 on Build GRCh38
rs371916556 Chr.X: 111295969-111295969 on Build GRCh38
rs1045402207 Chr.X: 111296281-111296281 on Build GRCh38
rs765069905 Chr.X: 111296054-111296054 on Build GRCh38
rs991287000 Chr.X: 111296232-111296232 on Build GRCh38
rs906259786 Chr.X: 111296014-111296014 on Build GRCh38

View Full Product Details