Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111687765-111688271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
UniGene
Hs.443061
Gene Chromosome Location
Chr.X: 111680741-111764164 on Build GRCh38
Transcripts
NM_001039210.4 NM_001099922.2 NM_001168385.2 NM_001257230.1 NM_001257231.1 NM_001257234.1 NM_001257235.2 NM_001257237.1 NM_001257239.2 NM_001257240.2 NM_001257241.2 NM_001324290.1 NM_001324291.1 NM_001324292.1 NM_001324293.1 NM_001324294.1 NM_018466.5 NR_033125.2 XM_006724693.2 XM_006724697.3 XM_006724698.3 XM_011531028.2 XM_011531030.1 XM_011531031.1 XM_011531032.1 XM_011531033.2 XM_017029846.1 XM_017029847.1 XM_017029848.1 XM_017029849.1 XM_017029850.1 XM_017029851.1

SNP Detail

SNP ID Chromosome Location
rs759406204 Chr.X: 111688029-111688029 on Build GRCh38
rs769458755 Chr.X: 111688235-111688235 on Build GRCh38
rs775557363 Chr.X: 111687939-111687939 on Build GRCh38
rs745739796 Chr.X: 111688214-111688214 on Build GRCh38
rs373095859 Chr.X: 111687942-111687942 on Build GRCh38
rs776425538 Chr.X: 111688017-111688017 on Build GRCh38
rs756431362 Chr.X: 111688007-111688007 on Build GRCh38
rs778025986 Chr.X: 111687808-111687808 on Build GRCh38
rs375827829 Chr.X: 111688032-111688032 on Build GRCh38
rs981755628 Chr.X: 111687874-111687874 on Build GRCh38
rs757970695 Chr.X: 111687866-111687866 on Build GRCh38
rs1039075198 Chr.X: 111688143-111688143 on Build GRCh38
rs781571236 Chr.X: 111687913-111687913 on Build GRCh38
rs771247315 Chr.X: 111687906-111687906 on Build GRCh38
rs887049356 Chr.X: 111688139-111688139 on Build GRCh38
rs1047603406 Chr.X: 111688096-111688096 on Build GRCh38
rs941331125 Chr.X: 111688153-111688153 on Build GRCh38
rs397518473 Chr.X: 111687924-111687925 on Build GRCh38
rs369113643 Chr.X: 111687852-111687852 on Build GRCh38
rs746970017 Chr.X: 111688162-111688162 on Build GRCh38
rs770214701 Chr.X: 111687930-111687930 on Build GRCh38
rs890388657 Chr.X: 111688117-111688117 on Build GRCh38
rs768715678 Chr.X: 111687950-111687950 on Build GRCh38
rs1000314512 Chr.X: 111688198-111688198 on Build GRCh38
rs780958214 Chr.X: 111688189-111688189 on Build GRCh38
rs866780677 Chr.X: 111687940-111687940 on Build GRCh38
rs777637325 Chr.X: 111687889-111687889 on Build GRCh38
rs771125470 Chr.X: 111688173-111688173 on Build GRCh38
rs1033101774 Chr.X: 111688228-111688228 on Build GRCh38
rs757232534 Chr.X: 111688172-111688172 on Build GRCh38
rs778570569 Chr.X: 111687840-111687840 on Build GRCh38
rs774606792 Chr.X: 111687957-111687957 on Build GRCh38
rs746639246 Chr.X: 111687898-111687898 on Build GRCh38
rs939234797 Chr.X: 111688109-111688109 on Build GRCh38
rs1008887637 Chr.X: 111688123-111688123 on Build GRCh38
rs767890935 Chr.X: 111687819-111687819 on Build GRCh38
rs777223517 Chr.X: 111688116-111688116 on Build GRCh38
rs899892654 Chr.X: 111688227-111688227 on Build GRCh38
rs765325071 Chr.X: 111688031-111688031 on Build GRCh38
rs757907272 Chr.X: 111688035-111688035 on Build GRCh38
rs752288318 Chr.X: 111688034-111688034 on Build GRCh38
rs746392733 Chr.X: 111687926-111687926 on Build GRCh38
rs926361487 Chr.X: 111687995-111687995 on Build GRCh38
rs762050093 Chr.X: 111687960-111687960 on Build GRCh38

View Full Product Details