Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111689296-111689835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
UniGene
Hs.443061
Gene Chromosome Location
Chr.X: 111680741-111764164 on Build GRCh38
Transcripts
NM_001099922.2 NM_001257230.1 NM_001257231.1 NM_001257234.1 NM_001257237.1 NM_001324292.1 NM_001324293.1 XM_006724693.2 XM_006724697.3 XM_006724698.3 XM_011531028.2 XM_011531030.1 XM_011531031.1 XM_011531032.1 XM_011531033.2 XM_011531034.2 XM_017029846.1 XM_017029847.1 XM_017029848.1 XM_017029849.1 XM_017029850.1 XM_017029851.1

SNP Detail

SNP ID Chromosome Location
rs773775095 Chr.X: 111689799-111689799 on Build GRCh38
rs973975665 Chr.X: 111689569-111689569 on Build GRCh38
rs193091810 Chr.X: 111689803-111689803 on Build GRCh38
rs190480604 Chr.X: 111689457-111689457 on Build GRCh38
rs765868604 Chr.X: 111689379-111689379 on Build GRCh38
rs867702773 Chr.X: 111689601-111689601 on Build GRCh38
rs145503272 Chr.X: 111689561-111689561 on Build GRCh38
rs149683115 Chr.X: 111689462-111689462 on Build GRCh38
rs965843918 Chr.X: 111689755-111689755 on Build GRCh38
rs759901066 Chr.X: 111689403-111689403 on Build GRCh38
rs538552280 Chr.X: 111689738-111689738 on Build GRCh38
rs932625184 Chr.X: 111689645-111689645 on Build GRCh38
rs974586564 Chr.X: 111689772-111689772 on Build GRCh38
rs753043451 Chr.X: 111689485-111689485 on Build GRCh38
rs1018697567 Chr.X: 111689730-111689730 on Build GRCh38
rs766788353 Chr.X: 111689808-111689808 on Build GRCh38
rs769042326 Chr.X: 111689476-111689476 on Build GRCh38
rs1035171270 Chr.X: 111689413-111689413 on Build GRCh38
rs771987402 Chr.X: 111689593-111689593 on Build GRCh38
rs950486954 Chr.X: 111689691-111689691 on Build GRCh38
rs908376341 Chr.X: 111689472-111689472 on Build GRCh38
rs921862887 Chr.X: 111689323-111689323 on Build GRCh38
rs983571054 Chr.X: 111689698-111689698 on Build GRCh38
rs946162934 Chr.X: 111689461-111689461 on Build GRCh38
rs941221602 Chr.X: 111689510-111689510 on Build GRCh38
rs907348357 Chr.X: 111689527-111689527 on Build GRCh38
rs969458146 Chr.X: 111689333-111689333 on Build GRCh38
rs980489091 Chr.X: 111689384-111689384 on Build GRCh38
rs1054893147 Chr.X: 111689389-111689389 on Build GRCh38
rs994502802 Chr.X: 111689594-111689594 on Build GRCh38
rs1051130024 Chr.X: 111689679-111689679 on Build GRCh38
rs921833166 Chr.X: 111689776-111689776 on Build GRCh38
rs754345449 Chr.X: 111689401-111689401 on Build GRCh38
rs898767291 Chr.X: 111689589-111689589 on Build GRCh38
rs774712259 Chr.X: 111689535-111689535 on Build GRCh38
rs372656175 Chr.X: 111689385-111689385 on Build GRCh38
rs778202475 Chr.X: 111689794-111689794 on Build GRCh38

View Full Product Details