Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111689519-111690014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
UniGene
Hs.443061
Gene Chromosome Location
Chr.X: 111680741-111764164 on Build GRCh38
Transcripts
NM_001099922.2 NM_001257230.1 NM_001257231.1 NM_001257234.1 NM_001257237.1 NM_001324292.1 NM_001324293.1 XM_006724693.2 XM_006724697.3 XM_006724698.3 XM_011531028.2 XM_011531030.1 XM_011531031.1 XM_011531032.1 XM_011531033.2 XM_011531034.2 XM_017029846.1 XM_017029847.1 XM_017029848.1 XM_017029849.1 XM_017029850.1 XM_017029851.1

SNP Detail

SNP ID Chromosome Location
rs773775095 Chr.X: 111689799-111689799 on Build GRCh38
rs973975665 Chr.X: 111689569-111689569 on Build GRCh38
rs913258916 Chr.X: 111689818-111689818 on Build GRCh38
rs193091810 Chr.X: 111689803-111689803 on Build GRCh38
rs906145608 Chr.X: 111689839-111689839 on Build GRCh38
rs867702773 Chr.X: 111689601-111689601 on Build GRCh38
rs1046194363 Chr.X: 111689855-111689855 on Build GRCh38
rs928743383 Chr.X: 111689900-111689900 on Build GRCh38
rs145503272 Chr.X: 111689561-111689561 on Build GRCh38
rs1045027261 Chr.X: 111689820-111689820 on Build GRCh38
rs965843918 Chr.X: 111689755-111689755 on Build GRCh38
rs932625184 Chr.X: 111689645-111689645 on Build GRCh38
rs538552280 Chr.X: 111689738-111689738 on Build GRCh38
rs974586564 Chr.X: 111689772-111689772 on Build GRCh38
rs58204668 Chr.X: 111689926-111689926 on Build GRCh38
rs1018697567 Chr.X: 111689730-111689730 on Build GRCh38
rs185442504 Chr.X: 111689890-111689890 on Build GRCh38
rs766788353 Chr.X: 111689808-111689808 on Build GRCh38
rs777027241 Chr.X: 111689841-111689841 on Build GRCh38
rs771987402 Chr.X: 111689593-111689593 on Build GRCh38
rs759862491 Chr.X: 111689912-111689912 on Build GRCh38
rs950486954 Chr.X: 111689691-111689691 on Build GRCh38
rs983571054 Chr.X: 111689698-111689698 on Build GRCh38
rs189001263 Chr.X: 111689948-111689948 on Build GRCh38
rs757420449 Chr.X: 111689840-111689840 on Build GRCh38
rs994502802 Chr.X: 111689594-111689594 on Build GRCh38
rs1051130024 Chr.X: 111689679-111689679 on Build GRCh38
rs921833166 Chr.X: 111689776-111689776 on Build GRCh38
rs898767291 Chr.X: 111689589-111689589 on Build GRCh38
rs778202475 Chr.X: 111689794-111689794 on Build GRCh38

View Full Product Details