Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 112775842-112776352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
UniGene
Hs.528051
Gene Chromosome Location
Chr.X: 112774877-112840946 on Build GRCh38
Transcripts
NM_001113490.1 NM_133265.2 XM_005262087.1 XM_005262090.1 XM_011530875.2 XM_017029289.1

SNP Detail

SNP ID Chromosome Location
rs988524814 Chr.X: 112776182-112776182 on Build GRCh38
rs1015351066 Chr.X: 112776116-112776116 on Build GRCh38
rs369130967 Chr.X: 112775991-112775991 on Build GRCh38
rs1016228322 Chr.X: 112775941-112775941 on Build GRCh38
rs35606486 Chr.X: 112775957-112775958 on Build GRCh38
rs955113638 Chr.X: 112776046-112776047 on Build GRCh38
rs968344930 Chr.X: 112776207-112776207 on Build GRCh38
rs989519166 Chr.X: 112776290-112776290 on Build GRCh38
rs895144156 Chr.X: 112776046-112776046 on Build GRCh38
rs980163844 Chr.X: 112776218-112776218 on Build GRCh38
rs1042912242 Chr.X: 112775888-112775888 on Build GRCh38
rs927257781 Chr.X: 112776279-112776279 on Build GRCh38
rs1031582428 Chr.X: 112776105-112776105 on Build GRCh38
rs763546169 Chr.X: 112776048-112776048 on Build GRCh38
rs753248969 Chr.X: 112776051-112776051 on Build GRCh38
rs998042462 Chr.X: 112776126-112776126 on Build GRCh38
rs1028236816 Chr.X: 112776128-112776128 on Build GRCh38
rs1049066603 Chr.X: 112776068-112776068 on Build GRCh38
rs138308883 Chr.X: 112776093-112776093 on Build GRCh38
rs779360256 Chr.X: 112775973-112775973 on Build GRCh38
rs780881608 Chr.X: 112776328-112776328 on Build GRCh38
rs549116776 Chr.X: 112775957-112775957 on Build GRCh38
rs901040154 Chr.X: 112776122-112776122 on Build GRCh38
rs1057000128 Chr.X: 112776319-112776319 on Build GRCh38
rs1004984195 Chr.X: 112776103-112776103 on Build GRCh38
rs41310719 Chr.X: 112776185-112776185 on Build GRCh38
rs758309366 Chr.X: 112775900-112775900 on Build GRCh38
rs945154455 Chr.X: 112775870-112775870 on Build GRCh38
rs1009089047 Chr.X: 112776089-112776091 on Build GRCh38
rs35667754 Chr.X: 112775877-112775878 on Build GRCh38
rs1029215521 Chr.X: 112776023-112776023 on Build GRCh38
rs996405315 Chr.X: 112775960-112775960 on Build GRCh38
rs1055407988 Chr.X: 112776012-112776012 on Build GRCh38

View Full Product Details