Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 116461605-116462113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT83
Entrez Gene ID
203413
Gene Name
cancer/testis antigen 83
Gene Aliases
CXorf61, KK-LC-1, KKLC1
UniGene
Hs.97892
Gene Chromosome Location
Chr.X: 116461686-116463033 on Build GRCh38
Transcripts
NM_001017978.3

SNP Detail

SNP ID Chromosome Location
rs990082155 Chr.X: 116461686-116461686 on Build GRCh38
rs782221085 Chr.X: 116461858-116461858 on Build GRCh38
rs918356171 Chr.X: 116461841-116461841 on Build GRCh38
rs782255184 Chr.X: 116462012-116462012 on Build GRCh38
rs782331654 Chr.X: 116462046-116462046 on Build GRCh38
rs781886365 Chr.X: 116461802-116461802 on Build GRCh38
rs781912973 Chr.X: 116461736-116461736 on Build GRCh38
rs782114367 Chr.X: 116461770-116461770 on Build GRCh38
rs901092623 Chr.X: 116462019-116462019 on Build GRCh38
rs782366893 Chr.X: 116461859-116461859 on Build GRCh38
rs199861269 Chr.X: 116461911-116461911 on Build GRCh38
rs782812378 Chr.X: 116461863-116461863 on Build GRCh38
rs781803198 Chr.X: 116461819-116461819 on Build GRCh38
rs148475461 Chr.X: 116462068-116462069 on Build GRCh38
rs782202522 Chr.X: 116461984-116461984 on Build GRCh38
rs782528472 Chr.X: 116461833-116461833 on Build GRCh38
rs782691513 Chr.X: 116461811-116461811 on Build GRCh38
rs782058480 Chr.X: 116461742-116461742 on Build GRCh38
rs12720093 Chr.X: 116461711-116461711 on Build GRCh38
rs782149102 Chr.X: 116461862-116461862 on Build GRCh38
rs782351401 Chr.X: 116461969-116461969 on Build GRCh38
rs782774785 Chr.X: 116461793-116461793 on Build GRCh38
rs782251724 Chr.X: 116461829-116461829 on Build GRCh38
rs781946735 Chr.X: 116461856-116461856 on Build GRCh38
rs782318584 Chr.X: 116461852-116461852 on Build GRCh38
rs188661465 Chr.X: 116461952-116461952 on Build GRCh38
rs148767220 Chr.X: 116461975-116461975 on Build GRCh38
rs782035972 Chr.X: 116462033-116462033 on Build GRCh38
rs781898960 Chr.X: 116461915-116461915 on Build GRCh38
rs781991937 Chr.X: 116461695-116461695 on Build GRCh38
rs781921711 Chr.X: 116461890-116461890 on Build GRCh38
rs782070523 Chr.X: 116461869-116461869 on Build GRCh38
rs267606323 Chr.X: 116461753-116461753 on Build GRCh38
rs201014112 Chr.X: 116462024-116462024 on Build GRCh38
rs782558701 Chr.X: 116461949-116461949 on Build GRCh38
rs782562201 Chr.X: 116461806-116461806 on Build GRCh38
rs782407719 Chr.X: 116461735-116461735 on Build GRCh38
rs367983468 Chr.X: 116462073-116462074 on Build GRCh38
rs370088796 Chr.X: 116461974-116461974 on Build GRCh38
rs782784212 Chr.X: 116461913-116461913 on Build GRCh38
rs375638675 Chr.X: 116461838-116461838 on Build GRCh38
rs782474980 Chr.X: 116461995-116461995 on Build GRCh38
rs928291853 Chr.X: 116461810-116461810 on Build GRCh38
rs149614986 Chr.X: 116461860-116461860 on Build GRCh38
rs782064385 Chr.X: 116461892-116461892 on Build GRCh38
rs916720834 Chr.X: 116461693-116461693 on Build GRCh38
rs372400858 Chr.X: 116461761-116461761 on Build GRCh38
rs782635564 Chr.X: 116462006-116462006 on Build GRCh38
rs782201405 Chr.X: 116461839-116461839 on Build GRCh38
rs781838860 Chr.X: 116461902-116461902 on Build GRCh38
rs201906931 Chr.X: 116461748-116461748 on Build GRCh38
rs999956164 Chr.X: 116462031-116462031 on Build GRCh38
rs1031541429 Chr.X: 116461652-116461652 on Build GRCh38
rs191793399 Chr.X: 116461701-116461701 on Build GRCh38
rs184008196 Chr.X: 116461893-116461893 on Build GRCh38
rs144361015 Chr.X: 116461924-116461924 on Build GRCh38
rs958126863 Chr.X: 116461666-116461666 on Build GRCh38

View Full Product Details