Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 118545182-118545702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
UniGene
Hs.368203
Gene Chromosome Location
Chr.X: 118495664-118686162 on Build GRCh38
Transcripts
NM_144658.3 XM_005262368.3 XM_005262369.4 XM_011531276.2

SNP Detail

SNP ID Chromosome Location
rs796271677 Chr.X: 118545282-118545282 on Build GRCh38
rs778959673 Chr.X: 118545299-118545299 on Build GRCh38
rs762168988 Chr.X: 118545403-118545403 on Build GRCh38
rs372685790 Chr.X: 118545438-118545438 on Build GRCh38
rs199613685 Chr.X: 118545271-118545271 on Build GRCh38
rs912788595 Chr.X: 118545585-118545585 on Build GRCh38
rs762553849 Chr.X: 118545371-118545371 on Build GRCh38
rs951654030 Chr.X: 118545243-118545245 on Build GRCh38
rs79880711 Chr.X: 118545281-118545282 on Build GRCh38
rs768026226 Chr.X: 118545274-118545274 on Build GRCh38
rs1047446264 Chr.X: 118545401-118545401 on Build GRCh38
rs763986360 Chr.X: 118545326-118545326 on Build GRCh38
rs958777808 Chr.X: 118545518-118545518 on Build GRCh38
rs1020969866 Chr.X: 118545589-118545589 on Build GRCh38
rs749519105 Chr.X: 118545294-118545294 on Build GRCh38
rs756254195 Chr.X: 118545282-118545282 on Build GRCh38
rs890376311 Chr.X: 118545556-118545556 on Build GRCh38
rs754721301 Chr.X: 118545422-118545423 on Build GRCh38
rs1010086413 Chr.X: 118545576-118545576 on Build GRCh38
rs990152318 Chr.X: 118545532-118545532 on Build GRCh38
rs370282294 Chr.X: 118545423-118545423 on Build GRCh38
rs753979516 Chr.X: 118545432-118545432 on Build GRCh38
rs187149324 Chr.X: 118545674-118545674 on Build GRCh38
rs142159478 Chr.X: 118545226-118545226 on Build GRCh38
rs979897964 Chr.X: 118545266-118545266 on Build GRCh38
rs761146274 Chr.X: 118545421-118545421 on Build GRCh38
rs751272805 Chr.X: 118545283-118545283 on Build GRCh38
rs55694052 Chr.X: 118545272-118545272 on Build GRCh38
rs866529893 Chr.X: 118545271-118545271 on Build GRCh38
rs151200381 Chr.X: 118545407-118545407 on Build GRCh38
rs914665814 Chr.X: 118545533-118545533 on Build GRCh38
rs181764263 Chr.X: 118545409-118545409 on Build GRCh38
rs1004562004 Chr.X: 118545261-118545263 on Build GRCh38
rs775136756 Chr.X: 118545249-118545249 on Build GRCh38
rs768387172 Chr.X: 118545376-118545376 on Build GRCh38
rs755192867 Chr.X: 118545297-118545297 on Build GRCh38
rs770796441 Chr.X: 118545315-118545315 on Build GRCh38
rs745799371 Chr.X: 118545352-118545352 on Build GRCh38
rs376163757 Chr.X: 118545251-118545253 on Build GRCh38
rs760489291 Chr.X: 118545263-118545263 on Build GRCh38
rs757677457 Chr.X: 118545426-118545427 on Build GRCh38
rs199603015 Chr.X: 118545247-118545249 on Build GRCh38
rs977467762 Chr.X: 118545584-118545585 on Build GRCh38
rs755504173 Chr.X: 118545359-118545359 on Build GRCh38
rs996000479 Chr.X: 118545535-118545535 on Build GRCh38
rs750984280 Chr.X: 118545416-118545416 on Build GRCh38
rs900247727 Chr.X: 118545462-118545462 on Build GRCh38
rs1030088586 Chr.X: 118545555-118545555 on Build GRCh38
rs377007516 Chr.X: 118545366-118545366 on Build GRCh38
rs750863872 Chr.X: 118545275-118545275 on Build GRCh38
rs979441320 Chr.X: 118545626-118545626 on Build GRCh38
rs780171655 Chr.X: 118545286-118545286 on Build GRCh38
rs746818414 Chr.X: 118545310-118545310 on Build GRCh38
rs754960498 Chr.X: 118545437-118545437 on Build GRCh38
rs138439558 Chr.X: 118545389-118545389 on Build GRCh38
rs967144725 Chr.X: 118545603-118545603 on Build GRCh38

View Full Product Details