Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120525261-120525762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4B
Entrez Gene ID
8450
Gene Name
cullin 4B
Gene Aliases
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2
UniGene
Hs.102914
Gene Chromosome Location
Chr.X: 120524589-120575829 on Build GRCh38
Transcripts
NM_001079872.1 NM_003588.3 XM_005262481.1 XM_006724784.1 XM_006724785.2 XM_011531399.2 XM_011531400.2 XM_011531401.1

SNP Detail

SNP ID Chromosome Location
rs751912130 Chr.X: 120525688-120525688 on Build GRCh38
rs759759999 Chr.X: 120525730-120525730 on Build GRCh38
rs376566357 Chr.X: 120525325-120525325 on Build GRCh38
rs910128097 Chr.X: 120525526-120525526 on Build GRCh38
rs985705420 Chr.X: 120525466-120525466 on Build GRCh38
rs764912137 Chr.X: 120525373-120525373 on Build GRCh38
rs976441292 Chr.X: 120525370-120525370 on Build GRCh38
rs941563824 Chr.X: 120525556-120525556 on Build GRCh38
rs190469409 Chr.X: 120525381-120525381 on Build GRCh38
rs939436186 Chr.X: 120525584-120525584 on Build GRCh38
rs748441588 Chr.X: 120525369-120525370 on Build GRCh38
rs895178178 Chr.X: 120525690-120525690 on Build GRCh38
rs932299609 Chr.X: 120525436-120525437 on Build GRCh38
rs1037203350 Chr.X: 120525561-120525561 on Build GRCh38
rs780536781 Chr.X: 120525601-120525601 on Build GRCh38
rs758177212 Chr.X: 120525457-120525457 on Build GRCh38
rs955822321 Chr.X: 120525299-120525299 on Build GRCh38
rs1031885242 Chr.X: 120525288-120525288 on Build GRCh38
rs897287922 Chr.X: 120525569-120525569 on Build GRCh38

View Full Product Details