Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120526428-120526923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4B
Entrez Gene ID
8450
Gene Name
cullin 4B
Gene Aliases
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2
UniGene
Hs.102914
Gene Chromosome Location
Chr.X: 120524589-120575829 on Build GRCh38
Transcripts
NM_001079872.1 NM_003588.3 XM_005262481.1 XM_006724784.1 XM_006724785.2 XM_011531399.2 XM_011531400.2 XM_011531401.1

SNP Detail

SNP ID Chromosome Location
rs770968413 Chr.X: 120526726-120526726 on Build GRCh38
rs751734223 Chr.X: 120526737-120526738 on Build GRCh38
rs767985470 Chr.X: 120526892-120526892 on Build GRCh38
rs143580749 Chr.X: 120526815-120526815 on Build GRCh38
rs763981937 Chr.X: 120526738-120526738 on Build GRCh38
rs748894655 Chr.X: 120526767-120526767 on Build GRCh38
rs769730471 Chr.X: 120526754-120526754 on Build GRCh38
rs926480197 Chr.X: 120526670-120526670 on Build GRCh38
rs773740692 Chr.X: 120526893-120526893 on Build GRCh38
rs1053424487 Chr.X: 120526690-120526690 on Build GRCh38
rs748316891 Chr.X: 120526721-120526721 on Build GRCh38
rs1043743337 Chr.X: 120526563-120526563 on Build GRCh38
rs760980494 Chr.X: 120526894-120526894 on Build GRCh38
rs185867059 Chr.X: 120526597-120526597 on Build GRCh38
rs1000134960 Chr.X: 120526658-120526658 on Build GRCh38
rs781476440 Chr.X: 120526764-120526764 on Build GRCh38
rs372169750 Chr.X: 120526732-120526732 on Build GRCh38
rs775146882 Chr.X: 120526760-120526760 on Build GRCh38
rs868818562 Chr.X: 120526826-120526826 on Build GRCh38
rs988160940 Chr.X: 120526548-120526548 on Build GRCh38
rs768424127 Chr.X: 120526797-120526797 on Build GRCh38
rs1057518598 Chr.X: 120526811-120526812 on Build GRCh38
rs191126833 Chr.X: 120526666-120526666 on Build GRCh38
rs745615241 Chr.X: 120526747-120526747 on Build GRCh38
rs761365165 Chr.X: 120526842-120526842 on Build GRCh38
rs762918169 Chr.X: 120526669-120526669 on Build GRCh38
rs939173103 Chr.X: 120526683-120526683 on Build GRCh38
rs1056173574 Chr.X: 120526696-120526696 on Build GRCh38
rs915191963 Chr.X: 120526582-120526582 on Build GRCh38
rs1048730072 Chr.X: 120526872-120526872 on Build GRCh38

View Full Product Details