Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120538568-120539077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4B
Entrez Gene ID
8450
Gene Name
cullin 4B
Gene Aliases
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2
UniGene
Hs.102914
Gene Chromosome Location
Chr.X: 120524589-120575829 on Build GRCh38
Transcripts
NM_001079872.1 NM_003588.3 XM_005262481.1 XM_006724784.1 XM_006724785.2 XM_011531399.2 XM_011531400.2 XM_011531401.1

SNP Detail

SNP ID Chromosome Location
rs1008466619 Chr.X: 120538945-120538945 on Build GRCh38
rs372205600 Chr.X: 120538965-120538965 on Build GRCh38
rs1021161653 Chr.X: 120538973-120538973 on Build GRCh38
rs772318747 Chr.X: 120538786-120538786 on Build GRCh38
rs748522493 Chr.X: 120538781-120538781 on Build GRCh38
rs948633195 Chr.X: 120538916-120538916 on Build GRCh38
rs767122119 Chr.X: 120538649-120538649 on Build GRCh38
rs1029977015 Chr.X: 120538878-120538878 on Build GRCh38
rs779335268 Chr.X: 120538778-120538778 on Build GRCh38
rs939345560 Chr.X: 120538874-120538874 on Build GRCh38
rs895728911 Chr.X: 120538883-120538887 on Build GRCh38
rs749827061 Chr.X: 120538773-120538773 on Build GRCh38
rs868357565 Chr.X: 120538668-120538668 on Build GRCh38
rs753692819 Chr.X: 120538996-120538996 on Build GRCh38
rs372816351 Chr.X: 120538746-120538746 on Build GRCh38
rs771007831 Chr.X: 120538818-120538818 on Build GRCh38
rs910023457 Chr.X: 120538676-120538676 on Build GRCh38
rs755854013 Chr.X: 120538880-120538880 on Build GRCh38
rs764004283 Chr.X: 120538964-120538964 on Build GRCh38
rs778293185 Chr.X: 120538779-120538780 on Build GRCh38
rs886310482 Chr.X: 120538603-120538603 on Build GRCh38
rs563431672 Chr.X: 120539040-120539040 on Build GRCh38
rs1047667855 Chr.X: 120538593-120538593 on Build GRCh38
rs751376524 Chr.X: 120538623-120538623 on Build GRCh38
rs979632819 Chr.X: 120538987-120538987 on Build GRCh38
rs775334942 Chr.X: 120538821-120538821 on Build GRCh38
rs972795719 Chr.X: 120538803-120538803 on Build GRCh38
rs941875129 Chr.X: 120538700-120538700 on Build GRCh38
rs996304926 Chr.X: 120538850-120538850 on Build GRCh38
rs797044862 Chr.X: 120538659-120538659 on Build GRCh38
rs967326118 Chr.X: 120538981-120538981 on Build GRCh38
rs747039580 Chr.X: 120538807-120538807 on Build GRCh38
rs959504366 Chr.X: 120539000-120539000 on Build GRCh38
rs751833365 Chr.X: 120538715-120538715 on Build GRCh38
rs949307023 Chr.X: 120538592-120538592 on Build GRCh38
rs755717522 Chr.X: 120538739-120538739 on Build GRCh38
rs991297232 Chr.X: 120539044-120539044 on Build GRCh38
rs755331312 Chr.X: 120538775-120538775 on Build GRCh38
rs773335186 Chr.X: 120538800-120538800 on Build GRCh38
rs904344842 Chr.X: 120538963-120538963 on Build GRCh38
rs757298030 Chr.X: 120538932-120538932 on Build GRCh38
rs756920132 Chr.X: 120538644-120538644 on Build GRCh38

View Full Product Details