Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120629805-120630303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1C1
Entrez Gene ID
29071
Gene Name
C1GALT1 specific chaperone 1
Gene Aliases
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS
UniGene
Hs.643920
Gene Chromosome Location
Chr.X: 120625674-120630150 on Build GRCh38
Transcripts
NM_001011551.2 NM_152692.4

SNP Detail

SNP ID Chromosome Location
rs759852579 Chr.X: 120630148-120630148 on Build GRCh38
rs768606749 Chr.X: 120629837-120629837 on Build GRCh38
rs935427424 Chr.X: 120629953-120629953 on Build GRCh38
rs892234150 Chr.X: 120630102-120630102 on Build GRCh38
rs753324870 Chr.X: 120630259-120630259 on Build GRCh38
rs1048204988 Chr.X: 120630079-120630079 on Build GRCh38
rs969977966 Chr.X: 120630195-120630195 on Build GRCh38
rs761859354 Chr.X: 120629950-120629950 on Build GRCh38
rs918284889 Chr.X: 120630254-120630254 on Build GRCh38
rs982964231 Chr.X: 120629923-120629923 on Build GRCh38
rs1045727107 Chr.X: 120629996-120629996 on Build GRCh38
rs932406033 Chr.X: 120629887-120629887 on Build GRCh38
rs374958394 Chr.X: 120630226-120630226 on Build GRCh38
rs141588084 Chr.X: 120630183-120630183 on Build GRCh38
rs981017742 Chr.X: 120630215-120630215 on Build GRCh38
rs111295814 Chr.X: 120629957-120629957 on Build GRCh38
rs180956880 Chr.X: 120629860-120629860 on Build GRCh38
rs753916084 Chr.X: 120630203-120630203 on Build GRCh38
rs929692543 Chr.X: 120630272-120630272 on Build GRCh38
rs772660860 Chr.X: 120629962-120629962 on Build GRCh38
rs926278690 Chr.X: 120630055-120630055 on Build GRCh38
rs1056719982 Chr.X: 120630250-120630250 on Build GRCh38
rs1021062843 Chr.X: 120630058-120630071 on Build GRCh38
rs1049434476 Chr.X: 120629892-120629892 on Build GRCh38
rs965355306 Chr.X: 120630122-120630122 on Build GRCh38
rs890863073 Chr.X: 120629903-120629903 on Build GRCh38
rs993915301 Chr.X: 120630072-120630072 on Build GRCh38
rs1056753661 Chr.X: 120630066-120630066 on Build GRCh38
rs777879851 Chr.X: 120630201-120630201 on Build GRCh38
rs964117504 Chr.X: 120629858-120629858 on Build GRCh38
rs956768732 Chr.X: 120630177-120630177 on Build GRCh38
rs890324951 Chr.X: 120630157-120630157 on Build GRCh38
rs1006786689 Chr.X: 120630101-120630101 on Build GRCh38
rs765993792 Chr.X: 120630114-120630114 on Build GRCh38
rs183531358 Chr.X: 120630071-120630071 on Build GRCh38
rs868608469 Chr.X: 120630208-120630208 on Build GRCh38
rs928193135 Chr.X: 120630232-120630232 on Build GRCh38
rs888284061 Chr.X: 120630089-120630089 on Build GRCh38
rs1027096987 Chr.X: 120629897-120629897 on Build GRCh38
rs751245167 Chr.X: 120630147-120630147 on Build GRCh38
rs377763257 Chr.X: 120629997-120629997 on Build GRCh38
rs3810744 Chr.X: 120630185-120630185 on Build GRCh38
rs905503475 Chr.X: 120630052-120630052 on Build GRCh38
rs948297251 Chr.X: 120629991-120629991 on Build GRCh38
rs939640473 Chr.X: 120630240-120630240 on Build GRCh38
rs767872882 Chr.X: 120630165-120630165 on Build GRCh38
rs1031905056 Chr.X: 120630105-120630105 on Build GRCh38
rs1008102633 Chr.X: 120629935-120629935 on Build GRCh38
rs749998292 Chr.X: 120629863-120629863 on Build GRCh38
rs938360086 Chr.X: 120630056-120630056 on Build GRCh38
rs915447632 Chr.X: 120629973-120629973 on Build GRCh38

View Full Product Details