Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 121048306-121048813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUD2
Entrez Gene ID
2747
Gene Name
glutamate dehydrogenase 2
Gene Aliases
GDH2, GLUDP1
UniGene
Hs.368538
Gene Chromosome Location
Chr.X: 121047608-121049942 on Build GRCh38
Transcripts
NM_012084.3

SNP Detail

SNP ID Chromosome Location
rs749503093 Chr.X: 121048764-121048764 on Build GRCh38
rs762633651 Chr.X: 121048633-121048633 on Build GRCh38
rs759066690 Chr.X: 121048427-121048427 on Build GRCh38
rs781372130 Chr.X: 121048538-121048538 on Build GRCh38
rs1055989264 Chr.X: 121048530-121048530 on Build GRCh38
rs374160903 Chr.X: 121048330-121048330 on Build GRCh38
rs763660196 Chr.X: 121048482-121048482 on Build GRCh38
rs779702832 Chr.X: 121048568-121048568 on Build GRCh38
rs374707988 Chr.X: 121048544-121048544 on Build GRCh38
rs1015893520 Chr.X: 121048675-121048675 on Build GRCh38
rs763572810 Chr.X: 121048638-121048638 on Build GRCh38
rs775117648 Chr.X: 121048627-121048627 on Build GRCh38
rs776965882 Chr.X: 121048582-121048582 on Build GRCh38
rs371555092 Chr.X: 121048695-121048695 on Build GRCh38
rs1025924529 Chr.X: 121048575-121048575 on Build GRCh38
rs770086797 Chr.X: 121048339-121048339 on Build GRCh38
rs895956907 Chr.X: 121048541-121048541 on Build GRCh38
rs368376259 Chr.X: 121048380-121048380 on Build GRCh38
rs756544884 Chr.X: 121048747-121048747 on Build GRCh38
rs759385556 Chr.X: 121048608-121048608 on Build GRCh38
rs1003134479 Chr.X: 121048515-121048515 on Build GRCh38
rs766069748 Chr.X: 121048377-121048377 on Build GRCh38
rs751112189 Chr.X: 121048648-121048648 on Build GRCh38
rs937401411 Chr.X: 121048511-121048511 on Build GRCh38
rs776219241 Chr.X: 121048587-121048587 on Build GRCh38
rs768917572 Chr.X: 121048360-121048360 on Build GRCh38
rs28928873 Chr.X: 121048504-121048504 on Build GRCh38
rs756234904 Chr.X: 121048550-121048550 on Build GRCh38
rs376591651 Chr.X: 121048711-121048711 on Build GRCh38
rs764871030 Chr.X: 121048432-121048432 on Build GRCh38
rs145381711 Chr.X: 121048503-121048503 on Build GRCh38
rs749591234 Chr.X: 121048567-121048567 on Build GRCh38
rs189203643 Chr.X: 121048670-121048670 on Build GRCh38
rs1043055660 Chr.X: 121048414-121048414 on Build GRCh38
rs746028809 Chr.X: 121048338-121048338 on Build GRCh38
rs1033987588 Chr.X: 121048521-121048521 on Build GRCh38
rs967231701 Chr.X: 121048645-121048645 on Build GRCh38
rs775646718 Chr.X: 121048340-121048340 on Build GRCh38
rs757831142 Chr.X: 121048517-121048517 on Build GRCh38
rs755277836 Chr.X: 121048782-121048782 on Build GRCh38
rs770928754 Chr.X: 121048579-121048579 on Build GRCh38
rs757888736 Chr.X: 121048472-121048472 on Build GRCh38
rs780412198 Chr.X: 121048557-121048557 on Build GRCh38
rs199542429 Chr.X: 121048756-121048756 on Build GRCh38
rs866972192 Chr.X: 121048774-121048774 on Build GRCh38
rs879031179 Chr.X: 121048364-121048364 on Build GRCh38
rs202048174 Chr.X: 121048571-121048571 on Build GRCh38
rs749394697 Chr.X: 121048357-121048357 on Build GRCh38
rs371442423 Chr.X: 121048396-121048396 on Build GRCh38
rs878905705 Chr.X: 121048392-121048392 on Build GRCh38
rs773281447 Chr.X: 121048375-121048375 on Build GRCh38
rs752142904 Chr.X: 121048451-121048451 on Build GRCh38
rs765072375 Chr.X: 121048612-121048612 on Build GRCh38
rs750951438 Chr.X: 121048714-121048714 on Build GRCh38

View Full Product Details