Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130012862-130013366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORL1
Entrez Gene ID
63035
Gene Name
BCL6 corepressor-like 1
Gene Aliases
BCoR-L1, CXorf10
UniGene
Hs.496748
Gene Chromosome Location
Chr.X: 129980285-130058083 on Build GRCh38
Transcripts
NM_001184772.2 NM_021946.4 XM_005262452.4 XM_005262453.4 XM_005262454.3 XM_005262455.4 XM_005262456.4 XM_006724776.3 XM_006724777.3 XM_006724779.2 XM_017029721.1 XM_017029722.1

SNP Detail

SNP ID Chromosome Location
rs973864978 Chr.X: 130012972-130012972 on Build GRCh38
rs779230116 Chr.X: 130013272-130013272 on Build GRCh38
rs750050005 Chr.X: 130013247-130013247 on Build GRCh38
rs779498199 Chr.X: 130012986-130012987 on Build GRCh38
rs750095557 Chr.X: 130013133-130013133 on Build GRCh38
rs745431187 Chr.X: 130013029-130013029 on Build GRCh38
rs761452039 Chr.X: 130013243-130013243 on Build GRCh38
rs368053264 Chr.X: 130013310-130013310 on Build GRCh38
rs760686487 Chr.X: 130013081-130013081 on Build GRCh38
rs760915291 Chr.X: 130013184-130013184 on Build GRCh38
rs748518635 Chr.X: 130013167-130013167 on Build GRCh38
rs751380690 Chr.X: 130013127-130013127 on Build GRCh38
rs141901231 Chr.X: 130012986-130012986 on Build GRCh38
rs770946442 Chr.X: 130013198-130013198 on Build GRCh38
rs755524355 Chr.X: 130013266-130013266 on Build GRCh38
rs753094647 Chr.X: 130012943-130012943 on Build GRCh38
rs766447653 Chr.X: 130013084-130013084 on Build GRCh38
rs756903033 Chr.X: 130013002-130013002 on Build GRCh38
rs534869537 Chr.X: 130013051-130013051 on Build GRCh38
rs761962656 Chr.X: 130012937-130012937 on Build GRCh38
rs764558968 Chr.X: 130012931-130012931 on Build GRCh38
rs755877513 Chr.X: 130013253-130013253 on Build GRCh38
rs149417325 Chr.X: 130013073-130013073 on Build GRCh38
rs4830173 Chr.X: 130013103-130013103 on Build GRCh38
rs368561000 Chr.X: 130013206-130013206 on Build GRCh38
rs925256170 Chr.X: 130013147-130013147 on Build GRCh38
rs372581512 Chr.X: 130013240-130013240 on Build GRCh38
rs779331157 Chr.X: 130013163-130013163 on Build GRCh38
rs367550651 Chr.X: 130012919-130012919 on Build GRCh38
rs200530257 Chr.X: 130013013-130013013 on Build GRCh38
rs755647755 Chr.X: 130013159-130013159 on Build GRCh38
rs145630843 Chr.X: 130013160-130013160 on Build GRCh38
rs774594848 Chr.X: 130012935-130012935 on Build GRCh38
rs775984166 Chr.X: 130012914-130012914 on Build GRCh38
rs774058034 Chr.X: 130013281-130013283 on Build GRCh38
rs200478600 Chr.X: 130013085-130013085 on Build GRCh38
rs373755234 Chr.X: 130012905-130012905 on Build GRCh38
rs376946512 Chr.X: 130013244-130013244 on Build GRCh38
rs981769962 Chr.X: 130013262-130013262 on Build GRCh38
rs36043572 Chr.X: 130013288-130013288 on Build GRCh38
rs779045172 Chr.X: 130013321-130013321 on Build GRCh38
rs766458162 Chr.X: 130013117-130013117 on Build GRCh38
rs375461860 Chr.X: 130013183-130013183 on Build GRCh38
rs768134462 Chr.X: 130013103-130013104 on Build GRCh38
rs756924604 Chr.X: 130013130-130013130 on Build GRCh38
rs147775035 Chr.X: 130013241-130013241 on Build GRCh38
rs770494128 Chr.X: 130012911-130012911 on Build GRCh38
rs368005608 Chr.X: 130013078-130013078 on Build GRCh38
rs754939100 Chr.X: 130012979-130012979 on Build GRCh38
rs185537912 Chr.X: 130013265-130013265 on Build GRCh38
rs748958709 Chr.X: 130013101-130013102 on Build GRCh38
rs41299086 Chr.X: 130013132-130013132 on Build GRCh38
rs772495890 Chr.X: 130013177-130013177 on Build GRCh38
rs772534414 Chr.X: 130013063-130013063 on Build GRCh38
rs370194529 Chr.X: 130012969-130012969 on Build GRCh38
rs1020902155 Chr.X: 130013010-130013010 on Build GRCh38
rs749788362 Chr.X: 130013162-130013162 on Build GRCh38
rs752641598 Chr.X: 130012981-130012981 on Build GRCh38
rs139887979 Chr.X: 130013052-130013052 on Build GRCh38
rs778220343 Chr.X: 130013304-130013304 on Build GRCh38
rs374936854 Chr.X: 130013075-130013075 on Build GRCh38
rs776734463 Chr.X: 130013202-130013202 on Build GRCh38
rs758119674 Chr.X: 130012982-130012982 on Build GRCh38
rs758649810 Chr.X: 130013298-130013298 on Build GRCh38
rs772248982 Chr.X: 130013077-130013077 on Build GRCh38
rs746653501 Chr.X: 130012989-130012989 on Build GRCh38
rs962418502 Chr.X: 130013210-130013210 on Build GRCh38
rs765213234 Chr.X: 130012980-130012980 on Build GRCh38
rs150631012 Chr.X: 130013042-130013042 on Build GRCh38
rs779677586 Chr.X: 130013263-130013263 on Build GRCh38

View Full Product Details